Get Up to $3,000 in BRANDTECH Products When You Spend $1,000 or More
When you buy $1,000 or more of qualifying BRANDTECH™ BRAND™ liquid handling products, VACUUBRAND™ vacuum pumps, or VITLAB™ labware, you’ll get up to $3,000 in extra BRANDTECH products at no additional cost:
- Spend $1,000, get $400
- Spend $3,000, get $1,500
- Spend $5,000, get $3,000
View qualifying and redeemable products below. After making your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
From BRAND liquid handling dispensers and pipettes to VACUUBRAND vacuum pumps and VITLAB labware, BRANDTECH has the products you need for your new lab.
Qualifying Products
BrandTech™ BRAND™ UV-Cuvettes - CERTIFIED LIFE SCIENCE QUALITY
BRAND™ disposable UV-cuvettes are a cost-effective, durable alternative to fragile quartz cuvettes in numerous applications
Disposable | Single-use |
---|---|
Height (Metric) | 45 mm |
Shape | Rectangle |
Path Length | 10 mm |
Lightpath | 10 mm |
Color | Clear |
Manufacturing Quality | CERTIFIED LIFE SCIENCE QUALITY |
Material | Plastic |
For Use With (Application) | DNA, RNA, Protein Analysis, Molecular Biology |
Width (Metric) | 12.5 mm |
Sterility | Non-sterile |
Includes | Cuvette Only |
For Use With (Equipment) | Spectrophotometers, Photometers |
Wavelength Range | 230 to 900 nm |
Pre-slit | No |
---|---|
Preservative | No |
Material | Polypropylene |
Closure Type | Snap Cap |
Autoclavable | Autoclavable |
Closure Material | LDPE |
Sterility | Non-sterile |
BRANDTECH™ VITLAB™ Polypropylene Griffin Beakers
Polypropylene Griffin Beakers are embossed with easily readable raised blue scale for accurate measurement.
Disposable | Reusable |
---|---|
Color | Translucent |
Style | Griffin |
Graduated | Yes |
Material | Polypropylene |
Certifications/Compliance | ISO 7056 |
Autoclavable | Autoclavable |
BRANDTECH™ VITLAB™ Graduated PMP Cylinders
VITLAB™ polymethylpentene graduated cylinders, certified Class A to DIN 12681 and ISO 6706
Type | Graduated Cylinder |
---|---|
Shape | Round |
Class | Class A |
Material | PMP |
Calibration | To Contain |
Certifications/Compliance | DIN 12681, ISO 6706 |
Autoclavable | Autoclavable |
Base Type | Hexagonal |
BRANDTECH™ VITLAB™ Wide-Mouth Reagent Bottles with Screw Caps
VITLAB wide mouth polypropylene reagent bottles with polypropylene screw caps have good chemical resistance. Autoclavable at 121 C for 20 minutes.
Product Type | Reagent Bottle |
---|---|
Mouth | Wide |
Color | Transparent |
Graduated | No |
Material | Polypropylene |
Closure Type | Screw Cap |
Autoclavable | Autoclavable |
Closure Material | Polypropylene |
Sterility | Non-sterile |
Closure Included | Yes |
Disposable | Reusable |
---|---|
Type | Beaker |
Temperature (English) Max. Operating | 302°F |
Color | Transparent |
Style | Griffin |
Graduated | Yes |
Scale | Red Graduated Scale |
Material | PMP |
Certifications/Compliance | ISO 7056 |
Temperature (Metric) Max. Operating | 150°C |
Autoclavable | Autoclavable |
For Use With (Application) | For Glass-Like Clarity |
BRANDTECH™ VITLAB™ Low Density Polyethylene Dropping Bottles
VITLAB Dropping bottles are made of low density polyethylene
Disposable | Reusable |
---|---|
Drop Size | Fine |
Shape | Round |
Product Type | Plastic Dropping Bottle |
Mouth | Narrow |
Color | Transparent |
Graduated | No |
Material | LDPE |
Closure Type | Screw Cap |
Autoclavable | Not Autoclavable |
Closure Material | PE-HD |
Sterility | Non-sterile |
Closure Included | Yes |
BRANDTECH™ VITLAB™ PFA Volumetric Flasks with Screw Caps
VITLAB™ PFA volumetric flasks are individually calibrated, and certified Class A to DIN EN ISO 1042.
Disposable | Reusable |
---|---|
Description | Individually calibrated ring-marking |
Mouth Style | Threaded |
Product Type | Volumetric Flask |
Graduated | No |
Temperature Range (English) | -328°F to +500°F |
Bottom Shape | Round |
Material | PFA |
Calibration | Calibrated To Contain |
Autoclavable | Autoclavable |
Temperature Range (Metric) | -200°C to +260°C |
Sterility | Non-sterile |
Closure Material | Perfluoxy Copolymer |
BRANDTECH™ VITLAB™ Class B PMP Volumetric Flasks with Screw Caps
An ideal alternative to glass flasks, these Class B PMP Volumetric Flasks with Screw Caps are designed for tight tolerances in the laboratory.
Disposable | Reusable |
---|---|
Mouth Style | Narrow |
Product Type | Volumetric Flask |
Color | Translucent |
Graduated | No |
Class | B |
Material | PMP |
Calibration | To Contain |
Closure Type | Screw |
Autoclavable | Autoclavable |
Temperature Range (Metric) | 121°C max. |
Sterility | Non-sterile |
Closure Material | Polypropylene |
BRANDTECH™ VITLAB™ POM or PMP Forceps
POM or PMP forceps offer resilience and flexibility. Available in various lengths and tip styles.
Product Type | Forceps |
---|---|
Style | Standard |
BRANDTECH™ VITLAB™ Styrene Acrylonitrile Copolymer Graduated Pitchers
VITLAB™ SAN graduated pitchers are transparent with molded graduations for durability
Disposable | Reusable |
---|---|
Shape | Round |
Product Type | Pouring Pitcher |
Mouth | Wide |
Graduated | Yes |
Material | SAN |
Autoclavable | Not Autoclavable |
Sterility | Non-sterile |
Closure Included | No |
BRANDTECH™ VITLAB™ Wide Mouth Bottles with Screw Caps
Transparent and resilient, with a wide mouth for easy filling.
Disposable | Single-use |
---|---|
Product Type | Storage and Transport Bottle |
Color | Transparent |
Quantity | 12/Pk. |
Material | LDPE |
Autoclavable | Not Autoclavable |
Sterility | Non-sterile |
BrandTech™ BRAND™ Amber Safety-Coated Bottles
Designed to protect light sensitive products and samples.Amber Safety-Coated Bottles are made of soda-lime glass with space-saving square design. Include polypropylene screw caps and LDPE pouring rings.
Disposable | Reusable |
---|---|
Includes | Polypropylene screw cap, LDPE pouring ring |
Operating Temperature | 80°C |
Shape | Square |
Product Type | Safety-coated Bottle |
Mouth | Narrow, Threaded |
Color | Amber |
Graduated | No |
Material | Soda Lime Glass |
Autoclavable | Not Autoclavable |
Sterility | Non-sterile |
Closure Included | Yes |
BRANDTECH™ VITLAB™ VITgrip™ Laboratory Bottles
Easy to hold laboratory bottles with accurate graduations
Disposable | Limited-use |
---|---|
Shape | Slim-waisted |
Closure Size | GL 45 |
Color | Clear |
Material | Polypropylene |
Closure Material | Polypropylene |
Sterility | Non-sterile |
Closure Included | Yes |
Product Type | Laboratory Bottle |
Mouth | Threaded |
Closure Liner | Unlined |
Graduated | Yes |
Closure Color | red |
Autoclavable | Autoclavable |