Learn More
BRANDTECH™ VITLAB™ Low Density Polyethylene Dropping Bottles
VITLAB Dropping bottles are made of low density polyethylene
$236.69 - $419.22
Specifications
Color Transparent
Material LDPE
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Thread Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Thread Size (Metric) Price Quantity & Availability  
14379055
View Documents Promotion Details
BRANDTECH™ VITLAB™
94587
50 mL GL 18
Pack of 24 Each for $283.31
Only null left
14379052
View Documents Promotion Details
BRANDTECH™ VITLAB™
94687
100 mL GL 18
Pack of 24 Each for $319.59
Only null left
14379053
View Documents Promotion Details
BRANDTECH™ VITLAB™
94987
250 mL GL 25
Pack of 12 Each for $236.69
Only null left
14379054
View Documents Promotion Details
BRANDTECH™ VITLAB™
95087
500 mL GL 25
Pack of 12 Each for $310.41
Only null left
14379056
View Documents Promotion Details
BRANDTECH™ VITLAB™
95187
1000 mL GL 32
Pack of 12 Each for $419.22
Only null left
Description
Description
  • Screw caps made of high density polyethylene.
  • Trim top to customize orifice size
  • Push-on cap helps secure contents
Specifications
Specifications
Transparent
Non-sterile
Reusable
No
Round
PE-HD
Not Autoclavable
LDPE
Yes
Fine
Narrow
Plastic Dropping Bottle
Screw Cap