Get 1 Extra Thermo Scientific Nalgene Product When You Buy 2
When you buy two identical qualifying Thermo Scientific™ Nalgene™ labware products, you’ll get one extra of the same product at no additional cost.
View qualifying and redeemable products below. Once you’ve made your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
Thermo Scientific Nalgene labware products are designed to be reusable and durable for long-term use in life sciences lab environments. These products are made from high-quality plastic materials that meet industry standards for pharmaceutical, laboratory, and food-grade applications and support safety and sustainability goals.
Qualifying Products
Thermo Scientific™ Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Carry out precise and efficient pouring with these low-form beakers, featuring a single-spout design and easy-to-read silk-screened graduations.
Disposable | Reusable |
---|---|
Type | Beaker |
ACT Label | Yes |
Color | Natural translucent |
Style | Griffin, Low Form |
Graduated | Yes |
Material | PPCO |
Certifications/Compliance | ISO/DIN 7056 |
Autoclavable | Autoclavable |
Thermo Scientific™ Nalgene™ Versi-Dry™ Surface Protectors
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Quickly absorb spills and cushion breakable labware with these surface protectors. Use as a bench protector, shelf liner or cabinet liner to protect against the harshest of chemicals.
Product Type | Surface Protector |
---|---|
Absorbancy | 750 mL/m2 |
Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Reduce breakage and chipping of glass pipette tips with the stackable and lightweight Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray.
Thermo Scientific™ Nalgene™ Large Polypropylene Sterilizing Pans
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Dry and sterilize instruments and labware with these large, polypropylene sterilizing pans, featuring rolled edges for extra stiffness and molded-in handles for easy carrying.
Type | Sterilizing Pan |
---|---|
Shape | Rectangular |
Description | Sterilizing pans are useful for drying and sterilizing instruments, labware; collecting and washing soiled lab equipment |
No. per Case | 6 |
Material | Polypropylene |
No. per Pack | 1 |
Autoclavable | Autoclavable |
Thermo Scientific™ Nalgene™ Polypropylene Economy Plastic Graduated Cylinders
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Dispense liquids easily with simple to read, molded-in graduations that won't rub off using these affordably-priced graduated cylinders.
Max. Temperature | 120°C |
---|---|
Type | Graduated Cylinder |
Shape | Round |
ACT Label | Yes |
Max. Temperature (English) | 248°F |
Material | Polypropylene |
Calibration | To Contain |
No. per Pack | 1 |
For Use With (Application) | Laboratories |
Base Type | Round |
Thermo Scientific™ Nalgene™ Acrylic Burets with PMP Tip, TFE Stopcock and Plug
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Accurately perform applications with these acrylic burets with PMP tip, TFE stopcock and plug, which meet ASTM E287 standards.
Type | Buret |
---|---|
No. per Case | 4 |
Class | Class B |
Material | Acrylic |
Certifications/Compliance | ASTME287 |
No. per Pack | 1 |
Thermo Scientific™ Nalgene™ Break-Resistant Self-Zeroing Automatic Burets
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Ensure repeatable, accurate results with these break-resistant, self-zeroing, automatic burets which meet ASTM E287.
Type | Automatic Buret |
---|---|
No. per Case | 4 |
Material | Acrylic |
Certifications/Compliance | ASTM E287 |
No. per Pack | 1 |
Graduations | 0.1 mL |
Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Transfer powders with these powder funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. A shatter-proof alternative to glass.
Material | Polypropylene |
---|---|
Autoclavable | Autoclavable |
Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Combine these flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.
Disposable | Reusable |
---|---|
Mouth Style | Narrow |
Bottom Shape | Plain |
Material | PPCO |
Autoclavable | Autoclavable |
Sterility | Non-sterile |
Thermo Scientific™ Nalgene™ Polypropylene Vacuum Flask
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Replace glass flasks with this flask, molded in one piece for full vacuum service with angled tubulation to prevent tipping.
No. per Case | 1 |
---|---|
Material | Polypropylene |
No. per Pack | 1 |
Autoclavable | Autoclavable |
Thermo Scientific™ Nalgene™ Class B Polypropylene Copolymer Volumetric Flasks with Closure
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Lighter in weight than glass, these flasks are break-resistant, offer excellent chemical resistance and protect against leakage and contamination.
Disposable | Reusable |
---|---|
Culture Area | Not applicable |
Mouth Style | Narrow |
Graduated | Yes |
Bottom Shape | Round |
Class | B |
Material | PPCO |
Certifications/Compliance | Meets ASTM E 288 and ISO 384 Class B tolerances for consistent quality and dependability |
Closure Type | Screw |
Autoclavable | Autoclavable but will affect accuracy |
Sterility | Non-sterile |
Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.
Material | Polypropylene |
---|
Thermo Scientific™ Nalgene™ Polypropylene Graduated Cylinders
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Conduct applications accurately with these graduated cylinders, which feature generous pour spouts and molded-in graduations.
Max. Temperature | 121°C |
---|---|
Type | Graduated Cylinder |
Material | Polypropylene |
Thermo Scientific™ Nalgene™ Super Versi-Dry™ Surface Protectors
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Absorb spills and cushion breakable items with long-lasting, waterproof, skid-resistant and chemical-resistant surface protectors.
Product Type | Surface Protector |
---|---|
Absorbancy | 1050 mL/m2 |
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Pans
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Repeatedly autoclave instruments and labware with these autoclavable polypropylene pans, featuring heavy-duty construction.
Description | Polypropylene Autoclavable Pan |
---|---|
Material | Polypropylene |
No. per Pack | 1 |
Autoclavable | Autoclavable |