Learn More
Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels
GREENER_CHOICE
Transfer powders with these powder funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. A shatter-proof alternative to glass.
$151.71 - $281.57
Specifications
Material Polypropylene
Autoclavable Autoclavable
View More Specs
Products 4
Catalog Number Mfr. No. Diameter (Metric)Top Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric)Top Capacity (Metric) Price Quantity & Availability  
10348A
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
42520065
64 mm 68 mL
Pack of 12 Each for $240.93
Case of 3 Pack for $722.79
Only null left
10348B
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
42520080
79mm 115 mL
Pack of 12 Each for $281.57
Case of 3 Pack for $844.71
Only null left
10348C
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
42520100
100 mm 243 mL
Pack of 6 Each for $180.71
Case of 4 Pack for $722.84
Only null left
10348D
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
42520150
145 mm 716 mL
Pack of 4 Each for $151.71
Case of 6 Pack for $910.26
Only null left
Description
Description
Transfer powders with Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.
  • Polypropylene is break-resistant and chemically compatible with a wide variety of lab reagents
  • Suitable for general lab use, it is a durable replacement for glass funnels
  • External ribs prevent airlocks
  • Autoclavable
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
Specifications
Polypropylene
Autoclavable