Get Extra Pipette Tips with Gilson Pipette Purchase
Buy any qualifying Gilson™ positive displacement pipette or pipette starter kit and get up to four packs of select Gilson pipette tips at no additional cost.
- Buy a Gilson™ PIPETMAN™, MyPIPETMAN™, or PIPETMAN L starter kit and get four packs of PIPETMAN Diamond Tips at no additional cost
- Buy a Gilson™ MICROMAN™ E positive displacement pipette and get one pack of Capillary Piston Tips at no additional cost
View the qualifying and redeemable products below. After making your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
For more than 60 years, Gilson has been creating pipettes with accuracy, comfort, and reliability in mind. Gilson offers a variety of pipettes in a range of ergonomic designs and volume sizes that enhance your pipetting experience and help you achieve reproducible, reliable results.
The MICROMAN E positive displacement pipette and capillary piston tips accurately measure viscous, volatile, foaming, hot, cold, or hazardous samples, and protect you from contamination.
Catalog Number | Description | Price | Quantity & Availability |
---|
Gilson™ PIPETMANTM TIPACKTM Racked Pipet Tips
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Engineered to be functional, convenient and robust. GilsonTM PIPETMANTM TIPACKTM Racked Pipet Tips are available in sterilized or autoclavable packages.
Disposable | Single-use |
---|---|
Tip Style | Standard |
Product Type | Pipet Tips |
Color | Clear |
Filtered | Non-filtered |
Material | Polypropylene |
Compatibility | Universal Fit |
Autoclavable | Autoclavable |
Gilson™ PIPETMANTM DIAMONDTM Blister Refill Filtered Pipette Tips
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
PIPETMANTM DiamondTM Performance Filter Tips: the quickest way to refill empty TIPACK boxes and Reload blocks for PIPETMAN pipetters.
Tip Style | Standard |
---|---|
Shape | Conical |
Color | Clear |
Quantity | 960 (10x96) |
Filtered | Filtered |
Fit for Purpose | Certified Quality |
Sterility | Sterile |
Clean Claims | DNA free, RNA free, RNAse free, DNAse free, ATP free, Endotoxin free |
Format | Rack |
For Use With (Equipment) | Gilson Pipetman |
Product Type | Pipette Tips |
Graduated | Yes |
Compatibility | PIPETMAN Pipettes |
Autoclavable | Autoclavable |
Product Line | Pipetman Diamond |
Gilson™ EXPERTTM Universal TOWERPACKTM Racked Pipette Tips Reload System
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Be green, save space: snaps tips into place in seconds with just one hand using the EXPERTTM Universal TOWERPACKTM Racked Pipette Tips Reload System.
Disposable | Single-use |
---|---|
Clean Claims | DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free |
Format | Refill |
For Use With (Equipment) | Universal Fit |
Product Type | Pipette Tips Reload System |
Graduated | Yes |
Filtered | Non-filtered |
Material | Inert Polymer |
Compatibility | Universal Fit |
Autoclavable | Not Autoclavable |
Fit for Purpose | Certified Quality |
Sterility | Non-sterile |
Product Line | EXPERT |
Gilson™ PIPETMANTM DIAMONDTM Tips Blister Refill, Filtered
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
PIPETMANTM DIAMONDTM Filtered Tips Blister Refill features individually wrapped racks of tips that are convenient, clean and eco-friendly.
Gilson™ EXPERTTM Universal TOWERPACKTM Racked Pipette Tips Reload Kit
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Be green, save space: snaps tips into place in seconds with just one hand with EXPERTTM Universal TOWERPACKTM Racked Pipette Tips Reload Kit plus TIPACK boxes.
Disposable | Single-use |
---|---|
Clean Claims | DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free |
Format | Refill |
For Use With (Equipment) | Universal Fit |
Product Type | Pipette Tips Reload Kit |
Graduated | Yes |
Filtered | Non-filtered |
Material | Inert Polymer |
Compatibility | Universal Fit |
Autoclavable | Autoclavable |
Fit for Purpose | Certified Quality |
Sterility | Non-sterile |
Product Line | EXPERT |
Gilson™ PIPETMANTM Diamond Blister Pack Refill Pipette Tips
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Autoclavable or gamma-ray sterlized
Product Type | Air Displacement Tips |
---|---|
Color | Clear |
Material | Polypropylene |
Compatibility | PIPETMAN Pipettes |
For Use With (Application) | Pipetting |
Product Line | Pipetman Diamond |
Gilson™ PIPETMANTM TipackTM Filter Tips
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Designed to meet the most stringent demands in various fields and applications such as PCR, molecular biology and cell technology. GilsonTM PIPETMANTM TipackTM Filter Tips are designed, developed, and manufactured in order to fit Gilson pipettes perfectly. They play a critical role in preventing aerosol-borne contamination.
Disposable | Single-use |
---|---|
Clean Claims | DNA free, RNA free, RNAse free, DNAse free, ATP free, Endotoxin free |
Tip Style | Standard |
Format | Rack |
Color | Clear |
Quantity | 960 (10x96) |
Filtered | Filtered |
Material | Polypropylene |
Compatibility | Universal Fit |
Autoclavable | Not Autoclavable |
Sterility | Sterile |
Product Line | Pipetman Tipack |
Gilson™ MICROMANTM E Capillary Pistons
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
MICROMANTM E Capillary Pistons provide reproducible, reliable results even with the most viscous or volatile liquids.
Disposable | Single-use |
---|---|
Tip Style | Positive Displacement |
For Use With (Equipment) | Gilson Microman |
Product Type | Capillary Piston |
Color | Clear |
Packaging | Box |
Filtered | Non-filtered |
Material | Polypropylene |
Compatibility | Microman E |
Gilson™ PIPETMANTM TOWERPACKTM Refill System
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Differentiates tip models with color-coded racks and indicates the compatible Gilson pipet models. GilsonTM PIPETMANTM TOWERPACKTM Refill System is available in autoclavable and sterilized versions.
Tip Style | Standard |
---|---|
Product Type | Tower Pack™ Pipet Refill System |
Color | Clear |
Filtered | Non-filtered |
Material | Polypropylene |
Compatibility | Universal Fit |
Autoclavable | Autoclavable |
Gilson™ DISTRITIPSTM Syringes
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Guarantees accuracy and precision. DISTRITIPSTM Syringes have an aliquot volume range from 1 μL to 1.25 mL.
Disposable | Single-use |
---|---|
Tip Style | Positive Displacement |
For Use With (Equipment) | DISTRIMAN Repeating Pipette |
Product Type | DistriTip™ Syringe |
Color | Clear |
Quantity | 50 Pk. |
Filtered | Non-filtered |
Material | Polypropylene |
Compatibility | Distriman |
For Use With (Application) | Viscous, volatile solutions |
Autoclavable | Autoclavable |
Product Line | DistriTips |
Gilson™ PIPETMANTM EcoPackTM Tips
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Ensures optimal performance of PIPETMAN pipettes across the full volume range. GilsonTM PIPETMANTM EcoPackTM Tips are fully autoclavable.
Disposable | Single-use |
---|---|
Tip Style | Universal |
Color | Clear |
Quantity | 10000 |
Filtered | Non-filtered |
Material | Polypropylene |
Sterility | Non-sterile |
Format | Bagged |
For Use With (Equipment) | Gilson Pipetman |
Product Type | Pipet Tips |
Graduated | Yes |
Compatibility | PIPETMAN Pipettes |
Autoclavable | Autoclavable |
Product Line | Pipetman EcoPack |
Gilson™ PIPETMANTM DIAMONDTM Blister Refill Non-Filtered Pipette Tips
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
PIPETMANTM DIAMOND Performance Tips: the quickest way to refill empty TIPACK boxes and Reload blocks for PIPETMAN pipettors.
Tip Style | Standard |
---|---|
For Use With (Equipment) | Gilson Pipetman |
Product Type | Pipette Tips |
Color | Clear |
Graduated | Yes |
Quantity | 960 (10x96) |
Filtered | Non-filtered |
Compatibility | PIPETMAN Pipettes |
Autoclavable | Autoclavable |
Fit for Purpose | Certified Quality |
Product Line | Pipetman Diamond |
Gilson™ PIPETMANTM EXPERTTM Tri-Fit Sterilized Filter Pipette Tips
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
PIPETMANTM EXPERTTM Sterilized Filter Pipette Tips features graduations at varying volumes allowing for quick-checks when aspirating samples.
Format | Rack |
---|---|
Color | Clear |
Packaging | Racked |
Graduated | Yes |
Filtered | Filtered |
Material | Polypropylene |
Sterility | Sterile |
Product Line | Pipetman EXPERT |
Gilson™ EXPERTTM Universal Fit Standard Pipette Tips
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Feature patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding while dispensing
Disposable | Single-use |
---|---|
Tip Style | Standard |
Shape | Conical |
Description | Standard Pipette Tips |
Color | Clear |
Filtered | Non-filtered |
Material | Polypropylene |
For Use With (Application) | Pipetting |
Clean Claims | RNase free, DNase free, Human DNA free, Endotoxins free, PCR Inhibitors free |
For Use With (Equipment) | Universal Fit |
Product Type | Air Displacement Tips |
Packaging | Racked |
Graduated | Yes |
Compatibility | Universal Fit |
Autoclavable | Autoclavable |
Product Line | EXPERT |
Gilson™ PIPETMANTM EXPERTTM Universal Fit Gel Loading Pipet Tips: Sterile
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Flat or round tip styles are available
Disposable | Single-use |
---|---|
Length (Metric) | 8.255cm |
Color | Clear |
Color Code | Yellow |
Length (English) | 12.75 in. |
Volume (Metric) | 1 to 200 μL |
Quantity | 800/Cs. |
Filtered | Non-filtered |
Material | Polypropylene |
For Use With (Application) | Gel Loading |
Sterility | Sterile |
Clean Claims | RNase free, DNase free, Human DNA free, Endotoxins free, PCR Inhibitors free |
Format | Rack |
For Use With (Equipment) | Universal Fit |
Product Type | Air Displacement Tips |
Packaging | Racked |
Model | Gel Loading |
Graduated | Yes |
Compatibility | Universal Fit |
Autoclavable | Autoclavable |
Graduations | 5μL |
Product Line | Pipetman EXPERT |