Gilson™ PIPETMAN™ DIAMOND™ Blister Refill Filtered Pipette Tips
PIPETMAN™ Diamond™ Performance Filter Tips: the quickest way to refill empty TIPACK boxes and Reload blocks for PIPETMAN pipetters.
$252.76 - $283.98
Specifications
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Rack
View More Specs
Products 6
Catalog Number Mfr. No. Volume (Metric) Model Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Model Price Quantity  
F172203G
View Documents Promotion Details
Gilson™
F172203
0.1 to 10 μL DFL10
Pack of 960 Each for $252.76
F172303G
View Documents Promotion Details
Gilson™
F172303
2 to 30 μL DFL30
Pack of 960 Each for $252.76
F172403G
View Documents Promotion Details
Gilson™
F172403
20 to 100 μL DFL100
Pack of 960 Each for $252.76
F172503G
View Documents Promotion Details
Gilson™
F172503
20 to 200 μL DFL200
Pack of 960 Each for $252.76
F172603G
View Documents Promotion Details
Gilson™
F172603
100 to 1000 μL DFL1000
Pack of 960 Each for $252.76
F172803G
View Documents Promotion Details
Gilson™
F172803
50 to 1200 μL DF1200ST
Pack of 960 Each for $283.98
Description
Description
  • Designed, developed and manufactured to fit Gilson pipetters perfectly.
  • Ensure optimal performance of your PIPETMAN pipetter across the full volume range.
  • Play a critical role in preventing aerosol-borne contamination.
  • Packaged in individually wrapped racks to ensure the best clearness and facilitate rack handling; top cover to handle sterilized pack.
  • Uses 45% less plastic than a regular rack box.
  • Fully autoclavable racks (moist heat, 121°C, 20 min., 1 bar); remove blister packaging before autoclaving.
  • Made of 100% pure virgin polypropylene; smooth hydrophobic surface for complete dispensing.
  • Two solid sealing rings provide a perfect seal
  • Ergonomic tip fitting: matching pipetter and tips reduces strain to fit and eject tips.
  • Fully traceable from mold to bench.
  • Graduated volume markers make it easy to visually check the volume aspirated.
  • Color-coded to differentiate tip models and indicate compatible Gilson pipetter models.
  • Available autoclavable or gamma-ray sterilized.
Sterilized Refills:
  • Sterilization date guaranteed for five years.
  • Irradiated by gamma-rays for more efficient sterilization.
  • Certified free of detectable DNA, RNA, DNase, RNase, ATP, Pyrogens and trace metals.
Order Info
10 Blister Refills of 96 racked tips
Specifications
Specifications
Filtered
Clear
Rack
Gilson Pipetman
Pipetman Diamond
Sterile
Autoclavable
Pipette Tips
Yes
Certified Quality
SDS
missing translation for 'documents'
missing translation for 'documents'