Learn More
Gilson™ PIPETMANTM TipackTM Filter Tips
GREENER_CHOICE
Designed to meet the most stringent demands in various fields and applications such as PCR, molecular biology and cell technology. GilsonTM PIPETMANTM TipackTM Filter Tips are designed, developed, and manufactured in order to fit Gilson pipettes perfectly. They play a critical role in preventing aerosol-borne contamination.
$251.82 - $251.82
Specifications
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Clean Claims DNA free, RNA free, RNAse free, DNAse free, ATP free, Endotoxin free
View More Specs
Products 8
Catalog Number Mfr. No. Length (Metric) Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Volume (Metric) Price Quantity  
F171103G
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F171103
3 cm 0.1 to 10 ÎĽL
Pack of 960 Each for $251.82
Only null left
F171203G
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F171203
4.5 cm 0.1 to 10 ÎĽL
Pack of 960 Each for $251.82
Only null left
F171303G Edge
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F171303
5.1 cm 2 to 30 ÎĽL
Pack of 960 Each for $251.82
Only null left
Edge
F171403G
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F171403
5.1 cm 10 to 100 ÎĽL
Pack of 960 Each for $251.82
Only null left
F171503G Edge
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F171503
5.1 cm 20 to 200 ÎĽL
Pack of 960 Each for $251.82
Only null left
Edge
F171603G
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F171603
5.1 cm 20 to 300 ÎĽL
Pack of 960 Each for $251.82
Only null left
F171703G
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F171703
5.1 cm 100 to 1000 ÎĽL
Pack of 960 Each for $251.82
Only null left
F171803G
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F171803
8.4 cm 100 to 1200 ÎĽL
Pack of 960 Each for $251.82
Only null left
Description
Description
  • Optimized geometry and pure polypropylene material ensure complete dispensing for consistent results.
  • Guarantees accuracy and precision.
  • No risk of contamination: Tips are manufactured in a clean room.
  • Gamma ray sterilized: Indicator dots used on Gilson's products undergo an irreversible color change upon sterilization.
  • Guaranteed free of DNA, RNA, DNase, RNase, ATP, Pyrogens and trace metals.
  • Full traceability from the mold to the bench guaranteed.
  • Easy identification with color-coded packaging.
Specifications
Specifications
Filtered
Clear
DNA free, RNA free, RNAse free, DNAse free, ATP free, Endotoxin free
Rack
960 (10x96)
Pipetman Tipack
Sterile
Not Autoclavable
Universal Fit
Standard
Single-use
Polypropylene
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'