missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis™ II Micro Balance, MCA User Interface
GREENER_CHOICE

Cubis™ II Micro Balance features fully customizable hardware, software, and connectivity

$42562.53 - $71896.02

Specifications

Certifications/Compliance 21 CFR PART 11 17025 USP 41
Display Type Color TFT Touchscreen
Type Micro Balance
Warranty 10 years
Voltage 100/240 V
View More Specs

Products 8
Catalog Number Mfr. No. Capacity Model Includes Price Quantity  
Catalog Number Mfr. No. Capacity Model Includes Price Quantity  
14561002
GREENER_CHOICE
View Documents
Sartorius
MCA66S2S00F
6.1 g Draft shield F: Filtered Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561001
GREENER_CHOICE
View Documents
Sartorius
MCA106S2S00F
10.1 g Draft shield F: Filtered Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561003
GREENER_CHOICE
View Documents
Sartorius
MCA36P2S00F
1.1 g, 2.1 g, 3.1 g Draft shield F: Filtered Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561006
GREENER_CHOICE
View Documents
Sartorius
MCA66S2S00M
6.1 g Draft shield M: Motorized Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561005
GREENER_CHOICE
View Documents
Sartorius
MCA106S2S00M
10.1 g Draft shield M: Motorized Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561223
GREENER_CHOICE
View Documents
Sartorius
MCA106SMS00
10.1 g Draft shield M: Motorized Balance, Accessories, Power supply, Manual, Factory test report, QP99 software N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561007
GREENER_CHOICE
View Documents
Sartorius
MCA36P2S00M
1.1 g, 2.1 g, 3.1 g Draft shield M: Motorized Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561243
GREENER_CHOICE
View Documents
Sartorius
MCA36PMS00
1.1 g, 2.1 g, 3.1 g Draft shield M: Motorized Balance, Accessories, Power supply, Manual, Factory test report, QP99 software N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • 21 CFR Part 11 compliance
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor

    (draft shield M), or gesture sensor (optional) learning capability

Warranty

10-year warranty

Specifications

Specifications

21 CFR PART 11 17025 USP 41
Micro Balance
100/240 V
Color TFT Touchscreen
10 years
Class I
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'