Get Up to $2,000 in Eppendorf Products with Purchase
When you spend $500, $1,000, or $2,000 on qualifying Eppendorf™ products, you’ll get a matching amount to spend on Eppendorf products at no additional cost. View qualifying and redeemable products below. After making your qualifying purchase, log in to the Redemption Center to redeem this offer.
- Spend $500, Get $500
- Spend $1,000, Get $1,000
- Spend $2,000, Get $2,000
Features and Benefits
For over 75 years, Eppendorf products have earned the trust of laboratory researchers. Start up your lab with the equipment, instruments, and consumables you need to complete your workflow, and enjoy more efficiency, semi-automation, and risk reduction with Eppendorf products.
Qualifying Products
Eppendorf™ Deepwell Plates 384-well, 200 μL
Eppendorf Deepwell plates offer efficient sample handling with features like OptiTrack™ matrix for rapid identification and g-Safe™ technology for stability during centrifugation. Safe Code option provides 3-level coding for safe sample and vessel identification.
Dimensions (L x W x H) | 127.8 x 85.5 x 25.1 mm |
---|---|
No. of Wells | 384 |
Well Color | Colorless |
Barcode | No Barcode |
Material | Polypropylene |
Volume (Metric) Well | 200 μL |
Autoclavable | Autoclavable |
Eppendorf™ Deepwell Plates 96-well, 500 μL
Eppendorf Deepwell plates offer efficient sample handling with features like OptiTrack™ matrix for rapid identification and g-Safe™ technology for stability during centrifugation. SafeCode variants provide secure three-level coding, including error correction for reliable identification, ensuring flexibility and accuracy in experimental workflows.
No. of Wells | 96 |
---|---|
Well Color | Colorless |
Barcode | No Barcode |
Material | Polypropylene |
Volume (Metric) Well | 500 μL |
Autoclavable | Autoclavable |
Eppendorf™ Protein LoBind™ Deepwell Plates
Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
Well Color | Colorless |
---|---|
Material | Polypropylene |
Max. RCF | 6000 x g |
Autoclavable | Autoclavable |
Sterility | Non-sterile |
Eppendorf™ Xplorer™ Electronic Pipette, Single Channel
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Eliminate potential error risks associated with manual pipetting with these pipettes. Eppendorf™ Xplorer™ Electronic Pipettes provide precisely adjustable parameters, reproducible and accurate results. Experience fatigue-free pipetting and maintain full control over pipetting procedures.
Type | Electronic Pipette |
---|---|
Number of Channels | 1 |
Product Line | Xplorer |
Eppendorf™ Xplorer™, Electronic Multi-Channel Pipette
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Eliminate potential error risks associated with manual pipetting with these pipettes. Eppendorf™ Xplorer™ Electronic Pipettes provide precisely adjustable parameters, reproducible and accurate results. Experience fatigue-free pipetting and maintain full control over pipetting procedures.
Type | Electronic Pipette |
---|---|
Product Line | Xplorer |
Eppendorf™ twin.tec™ Trace PCR Plates - PCR Plates
The Eppendorf twin.tec™ Trace PCR Plates feature an OptiTrack™ matrix and optical guiding grid for effortless well orientation and laser-engraved lot numbers for enhanced traceability.
Array | 8 x 12 |
---|---|
Skirt Style | Skirted |
No. of Wells | 96 |
Format | Standard |
Barcode | No Barcode |
Packaging | 5 Bags x 5 Plates |
Quantity | 25 Pk. |
Material | Polypropylene,Polycarbonate |
Volume (Metric) Well | 150 μL |
Fit for Purpose | PCR Ready |
Well Shape | Conical Bottom (V-well) |
Sterility | Non-sterile |
Product Line | Eppendorf |
Eppendorf™ Deepwell Plates 384-well, 200 μL
Eppendorf Deepwell plates offer efficient sample handling with features like OptiTrack™ matrix for rapid identification and g-Safe™ technology for stability during centrifugation. SafeCode variants provide secure three-level coding, including error correction for reliable identification, ensuring flexibility and accuracy in experimental workflows.
Eppendorf™ Research™ plus Mechanical Multi-Channel Pipette
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Adapt for specific liquids or other external conditions with these ultra-light fully autoclavable pipettes. Research™ plus Mechanical Multi-Channel Pipettes feature a spring loaded tip cone, a secondary adjustment option and an improved volume display.
Finger Style | Ergonomic |
---|---|
Type | Mechanical Pipette |
Autoclavable | Autoclavable |
Display | 4 Digits |
Eppendorf™ Xplorer™ Single-Channel Variable Pipette
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
From blue sky research to clinical applications, you’re working on challenging problems every day. You need the best tools and the best laboratory equipment, so when pipetting, the Eppendorf Xplorer or Xplorer plus electronic pipettes offer simplicity, precision, and reproducibility.
Disposable | Single-use |
---|---|
For Use With | Pipetting of Aqueous Liquids |
Plug Type | US |
Color | Red |
For Use With (Application) | Pipetting of Aqueous Liquids |
Battery Type | Lithium Polymer |
Type | Electronic Pipette |
Number of Channels | 1 |
Compatible Tips | Universal Tips, epT.I.P.S.™ |
Amperage | 1 A |
Increments | 0.02 μL |
Model | Single Channel Electronic |
Certifications/Compliance | Lot-specific Certificate |
Autoclavable | Autoclavable |
Product Line | Xplorer |
Eppendorf™ twin.tec™ real-time qPCR Plates, 96-well - qPCR Plates
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Eppendorf twin.tec™ real-time PCR plates, designed for low-volume qPCR, utilize white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.
Array | 8 x 12 |
---|---|
No. of Wells | 96 |
Format | Standard |
Well Color | White |
Product Type | PCR Plate |
Max. RCF | 6000 x g |
Autoclavable | Autoclavable |
Sterility | Non-sterile |
Rimmed | Yes |
Eppendorf™ twin.tec™ PCR Plates 96-well, 250 μL - PCR Plates
Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
Array | 8 x 12 |
---|---|
No. of Wells | 96 |
Format | Standard |
Product Type | PCR Plate |
Material | Polypropylene,Polycarbonate |
Volume (Metric) Well | 250 μL |
Autoclavable | Autoclavable |
Sterility | Non-sterile |
Eppendorf™ Deepwell Plates 96-well, 1000 μL
Eppendorf Deepwell Plates offer efficient sample handling with features like OptiTrack™ matrix for rapid identification and g-Safe™ technology for stability during centrifugation.
No. of Wells | 96 |
---|---|
Well Color | Colorless |
Barcode | No Barcode |
Material | Polypropylene |
Volume (Metric) Well | 1000 μL |
Autoclavable | Autoclavable |
Eppendorf™ Assay/Reader Microplate
Eppendorf Assay/Reader Microplates are designed for diverse microplate assays, optimizing absorbance, fluorescence, and chemiluminescence. With options for both top- and bottom-reading plate readers, they ensure clear signals even with low-concentration samples.
Purity or Quality Grade | PCR Clean (Non-sterile) |
---|---|
Max. RCF | 6000 x g |
Autoclavable | Autoclavable |
Eppendorf™ Microplate
Eppendorf Microplates provide exceptional stability during centrifugation with g-Safe™ technology and efficient sample identification with the OptiTrack™ Matrix. The RecoverMax™ well design and raised well rims ensure reliable sealing and facilitate maximum sample recovery.
Well Color | Colorless |
---|---|
Product Type | Microplate |
Barcode | No Barcode |
Material | Polypropylene |
Max. RCF | 6000 x g |
Autoclavable | Autoclavable |
Eppendorf™ twin.tec™ PCR Plates 96-well, 150 μL - PCR Plates
Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. Raised well rims enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
No. of Wells | 96 |
---|---|
Well Color | Colorless |
Material | Polypropylene,Polycarbonate |
Volume (Metric) Well | 150 μL |
Autoclavable | Autoclavable |
Sterility | Non-sterile |