missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Protein LoBind™ Deepwell Plates
Description
- Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
- High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
- Raised well rims and a smooth surface ensure reliable sealing in plates.
Compliance
IVD, SBS/ANSI™
Specifications
Specifications
| Certifications/Compliance | IVD, SBS/ANSI |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Autoclavable | Autoclavable |
| Packaging | 5 Bags x 8 Plates |
| Color | White |
| Material | Polypropylene |
| Volume (Metric) Well | 500 μL |
| No. of Wells | 96 |
| Show More |