Learn More
Eppendorf™ Xplorer™, Electronic Multi-Channel Pipette
Eliminate potential error risks associated with manual pipetting with these pipettes. Eppendorf™ Xplorer™ Electronic Pipettes provide precisely adjustable parameters, reproducible and accurate results. Experience fatigue-free pipetting and maintain full control over pipetting procedures.
$3353.10 - $4243.17
Specifications
Product Line Xplorer
Type Electronic Pipette
View More Specs
Products 8
Catalog Number Mfr. No. Volume (Metric) Number of Channels Color Increments Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Number of Channels Color Increments Price Quantity  
13-684-259
View Documents
Eppendorf™
4861000104
0.5 to 10 μL 8 Medium Gray 0.01 μL
Each for $3,353.10
Only null left
13-684-260
View Documents
Eppendorf™
4861000112
0.5 to 10 μL 12 Medium Gray 0.01 μL
Each for $4,137.19
Only null left
13-684-261
Accessories View Documents
Eppendorf™
4861000120
5 to 100 μL 8 Yellow 0.1 μL
Each for $3,353.10
Only null left
13-684-262
Accessories View Documents
Eppendorf™
4861000139
5 to 100 μL 12 Yellow 0.1 μL
Each for $4,243.17
Only null left
13-684-263
Accessories View Documents
Eppendorf™
4861000147
15 to 300 μL 8 Orange 0.2 μL
Each for $3,353.10
Only null left
13-684-264
Accessories View Documents
Eppendorf™
4861000155
15 to 300 μL 12 Orange 0.2 μL
Each for $4,243.17
Only null left
13-684-265
Accessories View Documents
Eppendorf™
4861000163
50 to 1200 μL 8 Green 1 μL
Each for $3,451.72
Only null left
05414055
View Documents
Eppendorf™
4861000171
50 to 1200 μL 12 Green 1 μL
Each for $3,534.18
Only null left
Description
Description
Eliminate potential error risks associated with manual pipetting with these pipettes. Eppendorf™ Xplorer™ Electronic Pipettes provide precisely adjustable parameters, reproducible and accurate results. Experience fatigue-free pipetting and maintain full control over pipetting procedures.
  • Electronic single, 8 and 12 channel pipettes designed to aspirate and dispense liquid volumes from 0.5 μL to 10 mL
  • Intuitive operating concept and ergonomic design
  • Nine different language settings: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish
  • Multifunctional rocker
  • Selection dial
  • Innovative ejector
  • Ergonomic display angle
  • Convenient handle
  • Powerful rechargeable battery
  • Flexible charging concept
  • Spring-loaded tip cone (except 5 and 10 mL)
  • Adjustment to external conditions (geographic height, liquids with high density or similar factors)
Specifications
Specifications
Xplorer
Electronic Pipette