missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ twin.tec™ PCR Plates 96-well, 150 μL - PCR Plates
Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. Raised well rims enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
$293.48 - $788.12
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Material | Polypropylene, Polycarbonate |
| Volume (Metric) Well | 150 μL |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Catalog Number | Mfr. No. | Color | Skirt Style | Product Type | Packaging | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Skirt Style | Product Type | Packaging | Price | Quantity & Availability | ||
|
E951020401
|
Eppendorf™
951020401 |
Colorless | Skirted | Standard PCR Plate | 5 Bags x 5 Plates |
Pack of 25 Each for $293.48
|
|
||
|
E951020427
|
Eppendorf™
951020427 |
Yellow | Skirted | Standard PCR Plate | 5 Bags x 5 Plates |
Pack of 25 Each for $293.48
|
|
||
|
E951020443
|
Eppendorf™
951020443 |
Green | Skirted | Standard PCR Plate | 5 Bags x 5 Plates |
Pack of 25 Each for $293.48
|
|
||
|
E951020460
|
Eppendorf™
951020460 |
Blue | Skirted | Standard PCR Plate | 5 Bags x 5 Plates |
Pack of 25 Each for $293.48
|
|
||
|
E951020486
|
Eppendorf™
951020486 |
Red | Skirted | Standard PCR Plate | 5 Bags x 5 Plates |
Pack of 25 Each for $293.48
|
|
||
|
361023833
|
Eppendorf™
0030129300 |
Colorless | Skirted | Microbiology PCR Plates | Individually Blister Wrapped |
Pack of 10 Each for $388.30
|
|
||
|
361023834
|
Eppendorf™
0030129318 |
Blue | Skirted | Microbiology PCR Plates | Individually Blister Wrapped |
Pack of 10 Each for $358.21
|
|
||
|
361023824
|
Eppendorf™
0030133307 |
Colorless | Unskirted | Low Profile PCR Plates | 5 Bags x 4 Plates |
Pack of 20 Each for $388.30
|
|
||
|
361023827
|
Eppendorf™
0030133331 |
Blue | Unskirted | Low Profile PCR Plates | 5 Bags x 4 Plates |
Pack of 20 Each for $358.21
|
|
||
|
361023829
|
Eppendorf™
0030133358 |
Colorless | Unskirted, Divisible | Divisible PCR Plates | 5 Bags x 4 Plates |
Pack of 20 Each for $788.12
|
|
||
|
361023830
|
Eppendorf™
0030133382 |
Blue | Unskirted, Divisible | Divisible PCR Plates | 5 Bags x 4 Plates |
Pack of 20 Each for $728.39
|
|
||
|
01671100
|
Eppendorf™
0030113560 |
Colorless | Skirted | PCR Plates | 5 Bags x 5 Plates |
Pack of 25 Each for $568.75
|
|
||
Description
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
- Exceptionally solid and torque-resistant polycarbonate frame.
- Improved well-to-well tolerance.
- Cut-away corner and alphanumeric grid-referencing.
- Raised well rims for effective sealing, also reduces risk of cross-contamination.
Certifications
Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.
Specifications
| Autoclavable | |
| 150 μL | |
| Non-sterile |
| Polypropylene, Polycarbonate | |
| 96 | |
| Colorless |