Get Up to $3,000 in BRANDTECH Products When You Spend $1,000 or More
When you buy $1,000 or more of qualifying BRANDTECH™ BRAND™ liquid handling products, VACUUBRAND™ vacuum pumps, or VITLAB™ labware, you’ll get up to $3,000 in extra BRANDTECH products at no additional cost:
- Spend $1,000, get $400
- Spend $3,000, get $1,500
- Spend $5,000, get $3,000
View qualifying and redeemable products below. After making your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
From BRAND liquid handling dispensers and pipettes to VACUUBRAND vacuum pumps and VITLAB labware, BRANDTECH has the products you need for your new lab.
Qualifying Products
BRANDTECH™ BOCHEM™ Support Jacks
Available in anodized aluminum (blue) or epoxy-powder-coated aluminum (green) decks
Product Type | Support Jack |
---|---|
For Use With (Application) | Fume hoods and marine and industrial laboratories, POM rollers |
BRANDTECH™ BOCHEM™ Stainless-Steel Support Jacks
Ideal for use in a wide variety of environments including fume hoods, solution baths, marine and industrial labs
Includes | Ratchet handle |
---|---|
Product Type | Support Jack |
Material | Stainless Steel, Epoxy Powder-coated Aluminum |
BrandTech™ BRAND™ seripettor™ Bottletop Dispensers
Suitable for use with bases, buffers, aqueous solutions of organic and inorganic substances, dilute non-oxidizing acids, and many polar solvents. Space-saving, low-rise design provides stability for easy one-handed operation and fit most common reagent bottles with the included adapters.
Warranty | 1 Year |
---|---|
Temperature (Metric) | 15°C to 40°C |
Color | Blue/Red/Clear |
Material | FEP,EPDM |
For Use With (Application) | Semi-automatic dispensing of alkaline, buffers, vitamin solutions, and nutrien media |
Density | Up to 2.2 g/cm3 |
Vapor Pressure | Up to 500 mbar |
Max. Pressure | 500 mbar |
Channels | 1 |
Includes | Discharge tube, telescoping filling tube, spare dispensing cartridge, 45/33mm and 45/38mm thread polypropylene adapters, operating manual |
Accuracy | ≤±1.2% |
Temperature (English) | 59°F to 104°F |
Angle | 90° |
Precision | ≤0.2% |
Product Type | Seripettor Bottletop Dispenser |
Autoclavable | Autoclavable |
Display | Analog |
BrandTech™ BRAND™ Macro Pipet Controller
Firmly holds and fills all volumetric and serologic pipets from 0.10 to 200mL. Macro Pipet Controller provides better airflow for effortless bellows compression. A single pipetting lever controls filling, meniscus setting, and discharge.
Grip Type | Bulb |
---|---|
Disposable | Reusable |
Type | Pipet Controller |
For Use With | Serologic Pipette |
Number of Channels | 1 |
For Use With (Equipment) | Serologic Pipettes |
Volume (Metric) | 0.1 to 200 mL |
Quantity | 1 Each |
Material | Polypropylene, Silicone Rubber |
Autoclavable | Autoclavable |
Adjustable Tip Spacing | No |
Sterility | Non-sterile |
BrandTech™ VACUUBRAND™ ME 8 NT Diaphragm Vacuum Pump
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
For a wide variety of non-corrosive and non-evaporative applications. ME-series pumps feature an ultimate vacuum of 52 Torr.
Enclosure Rating | IP40 |
---|---|
Temperature (Metric) Max | 40°C |
Overall Width (English) | 9.4 in. |
Temperature (Metric) Min. | 10°C |
Includes | Manual |
Flow Rate | 4.3/4.8 CFM |
Overall Height (English) | 7.7 in. |
Horsepower | 0.34 hp |
Overall Length (English) | 12.7 in. |
No. of Stages | 1 |
Overall Height (Metric) | 198.12 mm |
Weight (Metric) | 16.4 kg |
Temperature (English) Min | 50°F |
Temperature (English) Max. | 104°F |
Oil Type | Oil-Free |
Material | Aluminum Alloy housing/head cover, Aluminum Alloy diaphragm clamping disc, FPM diaphragm/valve, Aluminum Alloy connection tube, PBT Hose Nozzle, PVC Seal rings |
Pump Type | Diaphragm Pump |
Temperature (Metric) Storage | -10°C to +60°C |
Noise Level | 45 dbA |
Motor Speed | 150 to 1800 rpm |
Overall Width (Metric) | 23.8 cm |
Dimensions (L x W x H) | 12.7 x 9.4 x 7.7 in. (32.5 x 23.8 x 19.8 cm) |
Overall Length (Metric) | 325.12 mm |
Model | ME 8 NT |
Weight (English) | 36.1 lb. |
No. of Pump Heads | 4 |
Maximum Pressure | 16 psi (Inlet), 29 psi (Outlet) |
Max. Vacuum | 52 torr |
BrandTech™ VACUUBRAND™ ME 4C NT Dry Chemistry Diaphragm Pump
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Suitable for laboratory tasks such as vacuum filtration, degassing, solid phase extraction and many others
Enclosure Rating | IP40 |
---|---|
Overall Width (English) | 9.56 in. |
Includes | Manual |
Flow Rate | 2.3/2.6 CFM |
Overall Height (English) | 7.79 in. |
Horsepower | 0.24 hp |
Overall Length (English) | 10 in. |
No. of Stages | 1 |
Overall Height (Metric) | 198.12 mm |
Weight (Metric) | 11.1 kg |
Temperature (English) Min | 50°F |
Temperature (English) Max. | 104°F |
Applications | Filtration and Concentration of Solvents with Low Boiling Point |
Oil Type | Oil-Free |
Material | PTFE (Pump Head), Fluoroplastic (Case) |
Pump Type | Diaphragm Pump |
Noise Level | 45 dbA |
Motor Speed | 150 to 1800 rpm |
Overall Width (Metric) | 24.28 cm |
Overall Length (Metric) | 254 mm |
Model | ME 4C NT |
Frequency | 50/60 Hz |
Weight (English) | 24.47 lb. |
No. of Pump Heads | 2 |
Resistance Properties | Chemical-Resistant |
Maximum Pressure | 16 psi |
Max. Vacuum | 52.5 torr |
BrandTech™ VACUUBRAND™ MD 4C NT Dry Chemistry Diaphragm Vacuum Pump
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Suitable for many laboratory applications, including rotary evaporation, centrifugal concentration, gel drying, vacuum filtration, solid phase extraction and many other tasks
Enclosure Rating | IP40 |
---|---|
Temperature (Metric) Max | 40°C |
Overall Width (English) | 9.56 in. |
Temperature (Metric) Min. | 10°C |
Includes | Manual |
Flow Rate | 2/2.2 CFM |
Overall Height (English) | 7.7 in. |
Horsepower | 0.34 hp |
Overall Length (English) | 12.7 in. |
No. of Stages | 3 |
Overall Height (Metric) | 198.12 mm |
Weight (Metric) | 13.6 kg |
Temperature (English) Min | 50°F |
Temperature (English) Max. | 104°F |
Oil Type | Oil-Free |
Pump Type | Diaphragm Pump |
Temperature (Metric) Storage | -10°C to +60°C |
Noise Level | 45 dbA |
Motor Speed | 150 to 1800 rpm |
Overall Width (Metric) | 243.84 mm |
Overall Length (Metric) | 325.12 mm |
Model | MD 4C NT |
Frequency | 50/60 Hz |
Weight (English) | 29.9 lb. |
No. of Pump Heads | 4 |
Resistance Properties | Chemical-Resistant |
Maximum Pressure | 16 psi |
Max. Vacuum | 1.1 torr (without Gas Ballast), 2.3 torr (with Gas Ballast) |
BrandTech™ VACUUBRAND™ MD 4C NT + AK + EK Oil-free Diaphragm Vacuum Pump
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Designed for excellent corrosion resistance in routine biology and chemistry lab applications. The MD 4C NT + AK + EK Oil-free Diaphragm Vacuum Pump contains inlet/outlet catchpots, outlet condensers to enhance performance with condensable vapors to recover solvents.
Enclosure Rating | IP40 |
---|---|
Temperature (Metric) Max | 40°C |
Overall Width (English) | 9.56 in. |
Temperature (Metric) Min. | 10°C |
Includes | Manual |
Flow Rate | 2/2.2 CFM |
Overall Height (English) | 15.8 in. |
Horsepower | 0.34 hp |
Overall Length (English) | 12.8 in. |
No. of Stages | 3 |
Overall Height (Metric) | 401.32 mm |
Weight (Metric) | 17.3 kg |
Temperature (English) Min | 50°F |
Temperature (English) Max. | 104°F |
Oil Type | Oil-Free |
Pump Type | Diaphragm Pump |
Temperature (Metric) Storage | -10°C to +60°C |
Noise Level | 45 dbA |
Motor Speed | 150 to 1800 rpm |
Overall Width (Metric) | 243.84 mm |
Overall Length (Metric) | 325.12 mm |
Model | MD 4C NT + AK + EK |
Frequency | 50/60 Hz |
Weight (English) | 38.1 lb. |
No. of Pump Heads | 4 |
Resistance Properties | Chemical-Resistant |
Maximum Pressure | 16 psi |
Max. Vacuum | 1.1 torr (without Gas Ballast), 2.3 torr (with Gas Ballast) |
BrandTech™ VACUUBRAND™ PC 201 NT Dry Chemistry Vacuum System
VACUUBRAND PC 201 NT vacuum system. 230V, Continental Europe (CEE) plug
BrandTech™ VACUUBRAND™ ME 8C NT Chemistry Diaphragm Pump
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
VACUUBRAND Chemistry diaphragm pumps have fluoropolymer flowpaths for chemical resistance. Typical service intervals are in excess of 15,000 operating hours.
Enclosure Rating | IP40 |
---|---|
Temperature (Metric) Max | 40°C |
Overall Width (English) | 9.56 in. |
Temperature (Metric) Min. | 10°C |
Includes | Manual |
Flow Rate | 4/4.6 CFM |
Overall Height (English) | 7.7 in. |
Horsepower | 0.34 hp |
Overall Length (English) | 12.79 in. |
No. of Stages | 1 |
Overall Height (Metric) | 198.12 mm |
Weight (Metric) | 14.3 kg |
Temperature (English) Min | 50°F |
Temperature (English) Max. | 104°F |
Oil Type | Oil-Free |
Pump Type | Diaphragm Pump |
Temperature (Metric) Storage | -10°C to +60°C |
Noise Level | 45 dbA |
Motor Speed | 150 to 1800 rpm |
Overall Width (Metric) | 243.84 mm |
Overall Length (Metric) | 325.12 mm |
Model | ME 8C NT |
Weight (English) | 31.5 lb. |
No. of Pump Heads | 4 |
Resistance Properties | Chemical-Resistant |
Maximum Pressure | 16 psi |
Max. Vacuum | 52.5 torr (without Gas Ballast) |
BrandTech™ VACUUBRAND™ MZ 2 NT Diaphragm Vacuum Pumps
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
For a wide variety of non-corrosive and non-evaporative applications. MZ-series pumps have an ultimate vacuum of 5 Torr.
Enclosure Rating | IP40 |
---|---|
Temperature (Metric) Max | 40°C |
Overall Width (English) | 9.4 in. |
Temperature (Metric) Min. | 10°C |
Includes | Manual |
Flow Rate | 1.3/1.4 CFM |
Overall Height (English) | 7.7 in. |
Horsepower | 0.24 hp |
Overall Length (English) | 9.5 in. |
No. of Stages | 2 |
Overall Height (Metric) | 198.12 mm |
Weight (Metric) | 11 kg |
Temperature (English) Min | 50°F |
Temperature (English) Max. | 104°F |
Oil Type | Oil-Free |
Material | Aluminum Alloy housing/head cover, Aluminum Alloy diaphragm clamping disc, FPM diaphragm/valve, Aluminum Alloy connection tube, PBT Hose Nozzle, PVC Seal rings, Polyethylene tubing |
Pump Type | Diaphragm Pump |
Temperature (Metric) Storage | -10°C to +60°C |
Noise Level | 45 dbA |
Motor Speed | 150 to 1800 rpm |
Overall Width (Metric) | 23.8 cm |
Dimensions (L x W x H) | 9.5 x 9.4 x 7.7 in. (24.3 x 23.8 x 19.8 cm) |
Model | MZ 2 NT |
Frequency | 50/60 Hz |
Weight (English) | 24.25 lb. |
No. of Pump Heads | 2 |
Maximum Pressure | 16 psi (Inlet), 29 psi (Outlet) |
Max. Vacuum | 5.2 torr |
BRANDTECH™ VACUUBRAND™ Dry Chemistry Vacuum Systems
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Systems feature new generation NT-series vacuum pumps for high flow rates at working vacuum levels
Temperature (English) Min | 50°F |
---|---|
Enclosure Rating | IP40 |
Temperature (English) Max. | 104°F |
Electrical Requirements | 120 V 60 Hz |
Plug Type | US |
Temperature (Metric) Max | 40°C |
Material | PTFE (Pump Head) |
Overall Width (English) | 9.56 in. |
Temperature (Metric) Min. | 10°C |
Pump Type | Chemistry Diaphragm Pump |
Temperature (Metric) Storage | -10°C to +60°C |
Noise Level | 45 dbA |
Includes | One phase motor, on/off switch, cable with plug, operating manual |
Overall Height (English) | 15.82 in. |
Overall Width (Metric) | 243 mm |
Overall Length (Metric) | 326 mm |
Voltage | 100/115V, 120V |
Frequency | 50/60 Hz |
Overall Length (English) | 12.83 in. |
Certifications/Compliance | CE, DIN EN 1012-2:2011, DIN EN 12100:2001, DIN EN 50581:2013, DIN EN 61010-1:2011, DIN EN 61326-1:2013, IEC 61010-1:2010, NRTL Certification, TUV |
Resistance Properties | Chemical-Resistant |
Maximum Pressure | 1.1 bar |
Overall Height (Metric) | 402 mm |
BrandTech™ VACUUBRAND™ MD 4C NT+AK SYNCHRO + EK Dry Chemistry Vacuum System
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Provides simultaneous operation of two processes with only one pump.
BrandTech™ BRAND™ Dispensette™ S Trace Analysis Analog-adjustable Bottletop Dispensers
The Dispensette S Trace Analysis bottle dispenser is for precise volume dispensing of high-purity media for trace analysis.
Warranty | 2 Year |
---|---|
Temperature (Metric) | 15°C to 40°C |
Color | Gray |
Volume (Metric) | 1 to 10 mL |
For Use With (Application) | Dispensing acids, bases and hydrogen peroxide in the volume range from 1 to 10mL |
Density | Up to 3.8 g/cm3 |
Vapor Pressure | Up to 600 mbar |
Channels | 1 |
Accuracy | 0.5% (50ÎĽL) |
Temperature (English) | 59°F to 104°F |
Angle | 90° |
Precision | 0.1% (10ÎĽL) |
Product Type | Bottletop Dispenser |
Viscosity Rating | Up to 500 mm 2/s |
Increments | 0.2 mL |
Certifications/Compliance | ISO 9001 |
Autoclavable | Not Autoclavable |
Display | Analog |