missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ BRAND™ seripettor™ Bottletop Dispensers

Suitable for use with bases, buffers, aqueous solutions of organic and inorganic substances, dilute non-oxidizing acids, and many polar solvents. Space-saving, low-rise design provides stability for easy one-handed operation and fit most common reagent bottles with the included adapters.

$426.76 - $557.80

Specifications

Product Type Seripettor Bottletop Dispenser
Accuracy ≤±1.2%
Angle 90°
Autoclavable Autoclavable
Color Blue/Red/Clear
View More Specs

Includes: Bottle-top dispenser, for threaded bottles GL 45, includes discharge tube, filling tube, spare dispensing cartridge and PP adapters GL 32, GL 38 and S 40.

Products 3
Catalog Number Mfr. No. Volume (Metric) Increments Viscosity Rating Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Increments Viscosity Rating Price Quantity & Availability  
03840010
View Documents Promotion Details
BrandTech™ BRAND™
4720120
0.2 to 2 mL 0.04 mL 1000mm2/sec.
Each for $426.76
Only null left
 
13688246
View Documents Promotion Details
BrandTech™ BRAND™
4720140
1 to 10 mL 0.2 mL 150mm2/sec.
Each for $434.83
Only null left
 
13688247
View Documents Promotion Details
BrandTech™ BRAND™
4720150
2.5 to 25 mL 0.5 mL 75 mm2/sec.
Each for $557.80
Only null left
 
Description

Description

Suitable for use with bases, such as ammonia and sodium hydroxide; buffers; aqueous solutions of organic and inorganic substances; dilute non-oxidizing acids; and many polar solvents.

  • Fluid path consists of materials such as polyethylene, polypropylene and EPDM (ethylene-propylene-diene-rubber)
  • Full scale accuracy of ≤±1.2% and reproducibility of ≤0.2%
  • Fit most common reagent bottles with the included adapters
  • Space-saving, low-rise design provides stability for easy one-handed operation
  • With simple, clear volume adjustment for accuracy and a sturdy, supported discharge tube for safe operation
  • No fragile glass parts to break
  • Replaceable cylinder/piston cartridge has a wiping seal for renewability

This dispenser is not intended for use with scintillation fluid; non-polar solvents, such as xylene and most xylene substitutes; or strong, oxidizing or concentrated acids. For these liquids, choose the appropriate Dispensette™ bottletop dispenser.

Order Info

Unit disassembles in seconds for easy cleaning and maintenance. Replaceable piston and cylinder are permanently connected as a cartridge module for safety. If piston, cylinder, or seal are damaged, simply replace cartridge in seconds without tools.

Specifications

Specifications

Seripettor Bottletop Dispenser
90°
Blue/Red/Clear
1
Analog
Discharge tube, telescoping filling tube, spare dispensing cartridge, 45/33mm and 45/38mm thread polypropylene adapters, operating manual
≤0.2%
15°C to 40°C
1 Year
≤±1.2%
Autoclavable
Up to 2.2 g/cm3
FEP, EPDM
Semi-automatic dispensing of alkaline, buffers, vitamin solutions, and nutrien media
500 mbar
59°F to 104°F
Up to 500 mbar
Videos
SDS
Documents

Documents