Learn More
Cytiva Whatman™ Glass Microfiber Filter Paper Grade 6
Captures tiny particles without becoming weak. Whatman™ Glass Microfiber Filter Paper Grade 6 may also be used for determination of filterable substances and residue upon ignition or analysis of aggressive media.
$121.37 - $1578.66
Specifications
Material | Borosilicate Glass |
---|---|
Particle Retention | <1 ÎĽm |
Filtration Speed | 40 s/100mL/in2 |
Flow Rate | Slow |
Grade | GF 6 |
Catalog Number | Mfr. No. | Format | Diameter (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Format | Diameter (Metric) | Price | Quantity | |||||
09-924-256
|
Cytiva
10370002 |
Disc | 50 mm |
Pack of 200 Each for $414.47
|
|
|||||
09-924-258
|
Cytiva
10370004 |
Disc | 70 mm |
Pack of 100 Each for $245.21
|
|
|||||
09-924-257
|
Cytiva
10370003 |
Disc | 55 mm |
Pack of 100 Each for $190.44
|
|
|||||
09-924-259
|
Cytiva
10370005 |
Disc | 90 mm |
Pack of 100 Each for $336.35
|
|
|||||
09-924-260
|
Cytiva
10370006 |
Disc | 110 mm |
Pack of 100 Each for $465.46
|
|
|||||
09-924-261
|
Cytiva
10370007 |
Disc | 125 mm |
Pack of 100 Each for $579.39
|
|
|||||
09-924-262
|
Cytiva
10370008 |
Disc | 150 mm |
Pack of 100 Each for $787.71
|
|
|||||
09-924-263
|
Cytiva
10370010 |
Disc | 185 mm |
Pack of 100 Each for $882.11
|
|
|||||
09-924-265
|
Cytiva
10370012 |
Disc | 240 mm |
Pack of 100 Each for $1,151.19
|
|
|||||
09-924-264
|
Cytiva
10370011 |
Disc | 200 mm |
Pack of 100 Each for $1,016.64
|
|
|||||
09-924-268
|
Cytiva
10370020 |
Disc | 100 mm |
Pack of 100 Each for $531.66
|
|
|||||
09-924-267
|
Cytiva
10370019 |
Disc | 47 mm |
Pack of 200 Each for $186.24
|
|
|||||
09-924-266
|
Cytiva
10370018 |
Disc | 25 mm |
Pack of 200 Each for $121.37
|
|
|||||
09-924-269
|
Cytiva
10370050 |
Sheet |
Pack of 100 Each for $1,578.66
|
|
||||||
Description
Suitable for water and wastewater analysis, with extremely fine particle retention and added strength.- Sub micron filter retention efficiency to capture extremely small particles
- Inorganic binder for increased strength
- Temperature-resistant up to 500°C
- Can be used as a pre-filter for dense samples
Specifications
Borosilicate Glass | |
40 s/100mL/in2 | |
GF 6 |
<1 ÎĽm | |
Slow | |
350 ÎĽm |