Learn More
MilliporeSigma™ Isopore™ Membrane Filters, 0.4 μm
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
Supplier: MilliporeSigma™ HTBP04700
Description
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
- Environmental benefits include:
- Renewable Energy
Specifications
Not Autoclavable | |
Black | |
Polycarbonate | |
Non-sterile | |
Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence. | |
Hydrophilic | |
Filter Membranes | |
47 mm |
29 psi | |
Screen Filter | |
0.4 μm | |
Avg. 49 mL/min x cm2 (typical results @ 10 psi);18 mL/min x cm2/psi | |
10.0 L/min/cm2 | |
100/Pk. | |
Hydrophilic |