missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Isopore™ Membrane Filters, 0.4 μm

Catalog No. p-7232182
Change view
Click to view available options
Pore Size:
0.4 μm
1.2 μm
Diameter (Metric):
13 mm
25 mm
37 mm
47 mm
90 mm
142 mm
Color:
Black
White
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Diameter (Metric) Color
HTTP14250 0.4 μm 142 mm White
HTTP01300 0.4 μm 13 mm White
HTBP01300 0.4 μm 13 mm Black
RTTP01300 1.2 μm 13 mm White
HTTP02500 0.4 μm 25 mm White
HTBP02500 0.4 μm 25 mm Black
HTTP03700 0.4 μm 37 mm White
HTTP04700 0.4 μm 47 mm White
HTBP04700 0.4 μm 47 mm Black
HTTP09030 0.4 μm 90 mm White
RTTP14250 1.2 μm 142 mm White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. HTTP14250 Supplier MilliporeSigma™ Supplier No. HTTP14250

Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.

The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.

Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety

Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays

Specifications

Autoclavable Not Autoclavable
Bubble Point 29 psi
Color White
Filter Type Screen Filter
Material Polycarbonate
Pore Size 0.4 μm
Sterility Non-sterile
Flow Rate ≥50 mL/min/ cm2 (typical results @ 10 psi);18 mL/min x cm2/psi
For Use With (Application) Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence.
Gas Flow Rate 10.0 L/min/cm2
Hydrophobicity Hydrophilic
Quantity 50/Pk.
Type Filter Membranes
Wettability Hydrophilic
Diameter (Metric) 142 mm
Show More Show Less