Learn More
Sartorius Regenerated cellulose (RC) Membrane Filters
These solvent-resistant, hydrophilic membrane filters are excellently suited for particle removal from solvents.
Supplier:  Sartorius 1840747N
 View more versions of this product
Catalog No. 14-555-469
Only null left
Explore available promotions
Description
Description
These solvent-resistant, hydrophilic membrane filters are excellently suited for particle removal from solvents.
Regenerated cellulose membrane filters are reinforced with nonwoven cellulose. The membranes are mainly used for particle removal from solvents. For example, the 50mm diameter, 0.45μm pore size filter is standardly used to ultraclean and degas solvents and mobile phases for HPLC, in combination with an all-glass filter holder. Regenerated cellulose membrane also feature low non-specific-adsorption.
Specifications
Specifications
Autoclavable
Membrane Filter
Regenerated Cellulose
0.2 μm
For particle removal from solvents.
100/Pk.
Membrane Filter
Single-use
Disc
Cellulose
Non-sterile
Hydrophilic
160 to 200 μm
47 mm
Product Suggestions
Product Suggestions