Learn More
Promega Proteasome-GLO™ Cell-Based Assays
The Proteasome-Glo™ Cell-Based Assays are homogeneous, luminescent assays that measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in cultured cells.
Supplier: Promega G8860
Includes: Buffer, substrate and luciferin detection reagent
Description
- Homogeneous, Luminescent Assays that Measure Protease Activities in Cultured Cells
- Simple add-mix-measure protocol
- Chymotrypsin-Like, Trypsin-Like and Caspase-Like activity data directly from cellular environment
- Results within 10-30 minutes after reagent addition
Order Info
Assay system also available separately (Cat. No. PRG1200).
Specifications
Proteasome-Glo™ cell-based buffer | |
10 mL | |
−30 to −10°C |
Caspase-like cell-based assay kit | |
Proteasome-Glo™ cell-based Buffer, Z-nLPnLD-Glo™ Substrate and Luciferin detection reagent | |
Measures Chymotrypsin-like Protease activity in cultured cells |