missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Proteasome-GLO™ Cell-Based Assays
The Proteasome-Glo™ Cell-Based Assays are homogeneous, luminescent assays that measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in cultured cells.
$1123.91 - $3574.65
Specifications
| Buffer | Proteasome-Glo™ cell-based buffer |
|---|---|
| Product Type | Caspase-like cell-based assay kit |
| Includes | Proteasome-Glo™ cell-based Buffer, Z-nLPnLD-Glo™ Substrate and Luciferin detection reagent |
| Content And Storage | −30 to −10°C |
Includes: Buffer, substrate and luciferin detection reagent
Description
- Homogeneous, Luminescent Assays that Measure Protease Activities in Cultured Cells
- Simple add-mix-measure protocol
- Chymotrypsin-Like, Trypsin-Like and Caspase-Like activity data directly from cellular environment
- Results within 10-30 minutes after reagent addition
Order Info
Assay system also available separately (Cat. No. PRG1200).
Specifications
| Proteasome-Glo™ cell-based buffer | |
| Caspase-like cell-based assay kit | |
| Proteasome-Glo™ cell-based Buffer, Z-nLPnLD-Glo™ Substrate and Luciferin detection reagent | |
| −30 to −10°C |