Learn More
Promega™ HaloTag™ Interchangeable Labeling Technology
The HaloTag™ Technology enables covalent protein labeling and immobilization in vivo and in vitro.
Manufacturer: Promega™ G8251
Description
The HaloTag Technology is a platform technology for enabling covalent protein labeling and immobilization in vivo and in vitro. Products based on the HaloTag Technology enable researchers to study protein function in a biochemical and cellular environment. The technology is based on the efficient formation of a covalent bond between a specially designed reporter protein encoded by a HaloTag Vector and a specific ligand in living cells, in solution or on a solid support.
- Create a HaloTag fusion protein with one easy cloning step
- HaloTag protein creates strong and very specific covalent bonds with the different HaloTag ligands
- Cell membrane permeable fluorescent ligands allow imaging of live or fixed cells
- HaloTag PEG-Biotin Ligand for pull-down applications
Protein localization, Protein trafficking, Protein capture, Cell-to-gel analysis, Protein:protein interactions
Specifications
Protein localization, Protein trafficking, Protein capture, Cell to Gel analysis, Protein to Protein interactions | |
Fluorescent labels, affinity tags and attachments to a solid phase | |
30μL |
Fluid | |
HaloTag TMR Ligand | |
−30 to −10°C |
Promotions
-
Spend More, Get More on Promega Products
Purchase up to $1,000 in Promega reagents and get a matching amount of products at no additional cost.: 06/30/21