Promega HaloTag™ Interchangeable Labeling Technology DFS Item
The HaloTag™ Technology enables covalent protein labeling and immobilization in vivo and in vitro.
$531.87 - $916.26
Specifications
For Use With (Application) Protein localization, Protein trafficking, Protein capture, Cell to Gel analysis, Protein to Protein interactions
Includes Fluorescent labels, affinity tags and attachments to a solid phase
View More Specs
Products 11
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
PRG1002
View Documents
Promega
G1002
HaloTag Alexa Fluor 488 Ligand 15μL Each for $531.87
PRG2802
View Documents
Promega
G2802
HaloTag Oregon Green Ligand 15μL Each for $531.87
PRG3221
View Documents
Promega
G3221
HaloTag R110Direct Ligand 30μL Each for $916.26
PRG2991
View Documents
Promega
G2991
HaloTag TMRDirect Ligand 30μL Each for $916.26
PRG8252
View Documents
Promega
G8252
HaloTag TMR Ligand 15μL Each for $550.46
PRG8282
View Documents
Promega
G8282
HaloTag Biotin Ligand 15μL Each for $550.46
PRG8582
View Documents
Promega
G8582
HaloTag Coumarin Ligand 15μL Each for $550.46
PRG1001
View Documents
Promega
G1001
HaloTag Alexa Fluor 488 Ligand 30μL Each for $870.97
PRG2801
View Documents
Promega
G2801
HaloTag Oregon Green Ligand 30μL Each for $853.55
PR-G8251
View Documents
Promega
G8251
HaloTag TMR Ligand 30μL Each for $882.59
PRG8272
View Documents
Promega
G8272
HaloTag diAcFAM Ligand 30μL Each for $859.36
Description
Description
  • Covalently Attach Fluorescent Labels or Affinity Tags to Membrane or Intracellular Proteins
  • Cell permeant TMR, Oregon Green(R), diAcFAM and Coumarin ligands readily cross the cell membrane for labeling intracellular proteins
  • Cell impermeant tags include Alexa Fluor(R) 488 and 660 for quick cell surface labeling
Specifications
Specifications
Protein localization, Protein trafficking, Protein capture, Cell to Gel analysis, Protein to Protein interactions
Fluorescent labels, affinity tags and attachments to a solid phase