missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 3x DYKDDDDK Peptide
GREENER_CHOICE

Tricosa-peptide for competitively eluting DYKDDDK-tagged fusion proteins from immobilized supports

$770.79 - $2324.59

Specifications

Product Line Pierce™
Product Type Peptides
Protein Form Recombinant
Format Lyophilized Powder
Shipping Condition Wet Ice
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PIA36805
View Documents
Thermo Scientific™
A36805
5 mg
Each for $770.79
Only null left
 
PIA36806
View Documents
Thermo Scientific™
A36806
5 x 5 mg
Each for $2,324.59
Only null left
 
Description

Description

Pierce 3X DYKDDDDK Peptide is a lyophilized peptide with the sequence Met-Asp-Tyr-Lys-Asp-His-Asp-Gly-Asp-Tyr-Lys-Asp-His-Asp-Ile-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys (MDYKDHDGDYKDHDIDYKDDDDK). It has a molecular weight of 2861.9 g/mol. For use, reconstitute to 1.5 mg/mL in PBS.

This tricosa-peptide can be used to competitively elute DYKDDDDK-tagged fusion proteins from immobilized anti-DYKDDDDK affinity supports, especially when a DYKDDDDK-tagged protein is sensitive to low pH. At a 1.5 mg/mL concentration, complete release of tagged protein can be achieved in 5–10 minutes at ambient temperature.

Note: Pierce 3X DYKDDDDK Peptide may interfere with protein determination assays and absorbance at 280 nm. However, the peptide can be removed from the eluate by desalting (e.g., with Zeba Spin Desalting Columns, Cat. No. 89891).

Specifications

Specifications

Pierce™
Recombinant
Wet Ice
Lyophilized Powder
Peptides
Lyophilized Powder
Protein Purification and Isolation
Affinity Resin
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.