Learn More
MilliporeSigma™ Supelco™ Omnifit™ EZ Benchmark Columns with 1 Adjustable and 1 Fixed Endpiece
Omnifit™ Labware complete columns are supplied assembled and ready for use, with tubing and fittings to connect the column into an existing system.
$922.46 - $1773.88
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | |||||
111008373
|
MilliporeSigma™ Supelco™
K5100U |
50 mm | 6.6 mm |
Each for $1,173.75
|
|
|||||
111008783
|
MilliporeSigma™ Supelco™
K5102U |
150 mm | 6.6 mm |
Each for $922.46
|
|
|||||
111008723
|
MilliporeSigma™ Supelco™
K5103U |
250 mm | 6.6 mm |
Each for $979.22
|
|
|||||
111008691
|
MilliporeSigma™ Supelco™
K5104U |
330 mm | 6.6 mm |
Each for $939.14
|
|
|||||
111008640
|
MilliporeSigma™ Supelco™
K5105U |
400 mm | 6.6 mm |
Each for $985.00
|
|
|||||
111008641
|
MilliporeSigma™ Supelco™
K5106U |
100 mm | 10 mm |
Each for $985.00
|
|
|||||
111008616
|
MilliporeSigma™ Supelco™
K5107U |
150 mm | 10 mm |
Each for $1,007.22
|
|
|||||
111008577
|
MilliporeSigma™ Supelco™
K5108U |
250 mm | 10 mm |
Each for $1,047.07
|
|
|||||
111008549
|
MilliporeSigma™ Supelco™
K5109U |
330 mm | 10 mm |
Each for $1,066.53
|
|
|||||
111008407
|
MilliporeSigma™ Supelco™
K5111U |
500 mm | 10 mm |
Each for $1,160.03
|
|
|||||
111007942
|
MilliporeSigma™ Supelco™
K5112U |
100 mm | 15 mm |
Each for $1,301.57
|
|
|||||
111007939
|
MilliporeSigma™ Supelco™
K5114U |
250 mm | 15 mm |
Each for $1,452.85
|
|
|||||
111007455
|
MilliporeSigma™ Supelco™
K5117U |
500 mm | 15 mm |
Each for $1,529.51
|
|
|||||
111007749
|
MilliporeSigma™ Supelco™
K5116U |
400 mm | 15 mm |
Each for $1,394.08
|
|
|||||
111007442
|
MilliporeSigma™ Supelco™
K5119U |
150 mm | 25 mm |
Each for $1,555.66
|
|
|||||
111007290
|
MilliporeSigma™ Supelco™
K5122U |
400 mm | 25 mm |
Each for $1,636.85
|
|
|||||
111007091
|
MilliporeSigma™ Supelco™
K5123U |
500 mm | 25 mm |
Each for $1,773.88
|
|
|||||
Description
This configuration of Benchmark columns have one fixed endpiece as a bed support and one adjustable endpiece (plunger) to adjust the bed height (-AF). The adjustable endpiece is designed to give the user fine control over the movement of the endpiece, minimizing the risk of disturbance to the packed bed and giving 80 mm of height adjustment.Specifications
Omnifit | |
Chromatography |
Omnifit is a registered trademark of Diba Industries