missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Omnifit™ EZ Benchmark Columns with 1 Adjustable and 1 Fixed Endpiece
Description
This configuration of Benchmark columns have one fixed endpiece as a bed support and one adjustable endpiece (plunger) to adjust the bed height (-AF). The adjustable endpiece is designed to give the user fine control over the movement of the endpiece, minimizing the risk of disturbance to the packed bed and giving 80 mm of height adjustment.
Specifications
Specifications
| Diameter (Metric) | 6.6 mm |
| Length (Metric) | 50 mm |
| Product Line | Omnifit |
| Column Type | Chromatography |
Omnifit is a registered trademark of Diba Industries