Learn More
Labconco™ Fume Hood Accessory, SpillStopper™ Work Surface
SpillStopper Work Surface is designed for protecting your Labconco Protector™ Echo Filtered, Airo Filtered or XL Fume Hood from accidental spills.
$2235.37 - $14563.47
Specifications
Thickness (English) 1.25 in.
Depth (English) Cutout 0.4 in.
Depth (Metric) Cutout 1.0 cm
Product Type Work Surface
Thickness (Metric) 3.2 cm
View More Specs
Products 46
Catalog Number Mfr. No. Width (Metric) Cabinet Depth (Metric) Cabinet Includes For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Width (Metric) Cabinet Depth (Metric) Cabinet Includes For Use With (Equipment) Price Quantity  
13-118-391
View Documents
Labconco™
9500300
91.4 cm 76.2 cm No Cutouts Protector Echo Filtered Hood (36 in.)
Each for $2,235.37
Only null left
10-369-281
View Documents
Labconco™
9501500
152.4 cm 91.4 cm No Cutouts Protector Echo Filtered Hood (60 in.)
Each for $8,936.30
Only null left
10-369-263
View Documents
Labconco™
9501300
91.4 cm 91.4 cm No Cutouts Protector Echo Filtered Hood (36 in.)
Each for $5,962.87
Only null left
10-369-264
View Documents
Labconco™
9501301
91.4 cm 91.4 cm Right Rear (Cutout) Protector Echo Filtered Hood (36 in.)
Each for $6,682.27
Only null left
10-369-265
View Documents
Labconco™
9501302
91.4 cm 91.4 cm Right Side (Cutout) Protector Echo Filtered Hood (36 in.)
Each for $6,682.27
Only null left
10-369-266
View Documents
Labconco™
9501310
91.4 cm 91.4 cm Left Rear (Cutout) Protector Echo Filtered Hood (36 in.)
Each for $6,682.27
Only null left
10-369-267
View Documents
Labconco™
9501311
91.4 cm 91.4 cm Left, Right Rear (Cutout) Protector Echo Filtered Hood (36 in.)
Each for $7,353.67
Only null left
10-369-268
View Documents
Labconco™
9501312
91.4 cm 91.4 cm Left Rear, Right Side (Cutout) Protector Echo Filtered Hood (36 in.)
Each for $7,353.67
Only null left
10-369-269
View Documents
Labconco™
9501320
91.4 cm 91.4 cm Left Side (Cutout) Protector Echo Filtered Hood (36 in.)
Each for $3,286.36
Only null left
10-369-270
View Documents
Labconco™
9501321
91.4 cm 91.4 cm Left Side, Right Rear (Cutout) Protector Echo Filtered Hood (36 in.)
Each for $3,616.56
Only null left
10-369-271
View Documents
Labconco™
9501322
91.4 cm 91.4 cm Work Surface Protector Echo Filtered Hood (36 in.)
Each for $3,616.56
Only null left
10-369-272
View Documents
Labconco™
9501400
121.9 cm 91.4 cm No Cutouts Protector Echo Filtered Hood (48 in.)
Each for $7,465.57
Only null left
10-369-273
View Documents
Labconco™
9501401
121.9 cm 91.4 cm Right Rear (Cutout) Protector Echo Filtered Hood (48 in.)
Each for $8,137.00
Only null left
10-369-274
View Documents
Labconco™
9501402
121.9 cm 91.4 cm Right Side (Cutout) Protector Echo Filtered Hood (48 in.)
Each for $8,137.00
Only null left
10-369-275
View Documents
Labconco™
9501410
121.9 cm 91.4 cm Left Rear (Cutout) Protector Echo Filtered Hood (48 in.)
Each for $8,137.00
Only null left
10-369-276
View Documents
Labconco™
9501411
121.9 cm 91.4 cm Left, Right Rear (Cutout) Protector Echo Filtered Hood (48 in.)
Each for $8,840.40
Only null left
10-369-277
View Documents
Labconco™
9501412
121.9 cm 91.4 cm Left Rear, Right Side (Cutout) Protector Echo Filtered Hood (48 in.)
Each for $8,840.40
Only null left
10-369-278
View Documents
Labconco™
9501420
121.9 cm 91.4 cm Left Side (Cutout) Protector Echo Filtered Hood (48 in.)
Each for $8,137.00
Only null left
10-369-279
View Documents
Labconco™
9501421
121.9 cm 91.4 cm Left Side, Right Rear (Cutout) Protector Echo Filtered Hood (48 in.)
Each for $8,840.40
Only null left
10-369-280
View Documents
Labconco™
9501422
121.9 cm 91.4 cm Left, Right Side (Cutout) Protector Echo Filtered Hood (48 in.)
Each for $8,840.40
Only null left
10-369-290
View Documents
Labconco™
9501600
182.9 cm 91.4 cm No Cutouts Protector Echo Filtered Hood (72 in.)
Each for $5,126.08
Only null left
10-369-291
View Documents
Labconco™
9501601
182.9 cm 91.4 cm Right Rear (Cutout) Protector Echo Filtered Hood (72 in.)
Each for $5,456.30
Only null left
10-369-292
View Documents
Labconco™
9501602
182.9 cm 91.4 cm Right Side (Cutout) Protector Echo Filtered Hood (72 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-293
View Documents
Labconco™
9501610
182.9 cm 91.4 cm Left Rear (Cutout) Protector Echo Filtered Hood (72 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-294
View Documents
Labconco™
9501611
182.9 cm 91.4 cm Left, Right Rear (Cutout) Protector Echo Filtered Hood (72 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-295
View Documents
Labconco™
9501612
182.9 cm 91.4 cm Left Rear, Right Side (Cutout) Protector Echo Filtered Hood (72 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-296
View Documents
Labconco™
9501620
182.9 cm 91.4 cm Left Side (Cutout) Protector Echo Filtered Hood (72 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-297
View Documents
Labconco™
9501621
182.9 cm 91.4 cm Left Side, Right Rear (Cutout) Protector Echo Filtered Hood (72 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-298
View Documents
Labconco™
9501622
182.9 cm 91.4 cm Left, Right Side (Cutout) Protector Echo Filtered Hood (72 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-299
View Documents
Labconco™
9501800
243.8 cm 91.4 cm No Cutouts Protector Echo Filtered Hood (96 in.)
Each for $6,509.80
Only null left
10-369-300
View Documents
Labconco™
9501801
243.8 cm 91.4 cm Right Rear (Cutout) Protector Echo Filtered Hood (96 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-301
View Documents
Labconco™
9501802
243.8 cm 91.4 cm Right Side (Cutout) Protector Echo Filtered Hood (96 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-302
View Documents
Labconco™
9501810
243.8 cm 91.4 cm Left Rear (Cutout) Protector Echo Filtered Hood (96 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-303
View Documents
Labconco™
9501811
243.8 cm 91.4 cm Left, Right Rear (Cutout) Protector Echo Filtered Hood (96 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-304
View Documents
Labconco™
9501812
243.8 cm 91.4 cm Left Rear, Right Rear Protector Echo Filtered Hood (96 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-305
View Documents
Labconco™
9501820
243.8 cm 91.4 cm Left Side Protector Echo Filtered Hood (96 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-306
View Documents
Labconco™
9501821
243.8 cm 91.4 cm Left Side, Right Rear Protector Echo Filtered Hood (96 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-307
View Documents
Labconco™
9501822
243.8 cm 91.4 cm Left, Right Side Protector Echo Filtered Hood (96 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-282
View Documents
Labconco™
9501501
152.4 cm 91.4 cm Right Rear (Cutout) Protector Echo Filtered Hood (60 in.)
Each for $9,623.73
Only null left
10-369-283
View Documents
Labconco™
9501502
152.4 cm 91.4 cm Right Side (Cutout) Protector Echo Filtered Hood (60 in.)
Each for $9,623.73
Only null left
10-369-284
View Documents
Labconco™
9501510
152.4 cm 91.4 cm Left Rearf (Cutout) Protector Echo Filtered Hood (60 in.)
Each for $9,623.73
Only null left
10-369-285
View Documents
Labconco™
9501511
152.4 cm 91.4 cm Left, Right Rear (Cutout) Protector Echo Filtered Hood (60 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-286
View Documents
Labconco™
9501512
152.4 cm 91.4 cm Left Rear, Right Side (Cutout) Protector Echo Filtered Hood (60 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-287
View Documents
Labconco™
9501520
152.4 cm 91.4 cm Left Side (Cutout) Protector Echo Filtered Hood (60 in.)
Each for $9,623.73
Only null left
10-369-288
View Documents
Labconco™
9501521
152.4 cm 91.4 cm Left Side, Right Rear (Cutout) Protector Echo Filtered Hood (60 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-369-289
View Documents
Labconco™
9501522
152.4 cm 91.4 cm Left, Right Side (Cutout) Protector Echo Filtered Hood (60 in.) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Molded from special formulation of corrosion-resistant epoxy resin
  • Dished surface to contain spills
  • Contoured to conform to interior liner of Protector Echo, Airo and XL hoods
  • Pre-drilled 5.8cm (1.5 in.) diameter holes for venting
  • Pre-drilled 6.4cm (2.5 in.) for plumbing pass-through
  • Front edge has large radius to aerodynamically direct airflow into hood
  • May be ordered with a pre-cut 15.2 x 7.6cm (6 x 3 in.) oval cupsink cutout(s) in left rear, right rear, left side, or right side
  • Cupsink sold separately
Warranty
One year
Specifications
Specifications
1.25 in.
1.0 cm
3.2 cm
0.4 in.
Work Surface
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'