Learn More
MilliporeSigma™ Isopore™ Membrane Filters, 0.4 μm
Description
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Bubble Point | 29 psi |
| Color | Black |
| Filter Type | Screen Filter |
| Material | Polycarbonate |
| Pore Size | 0.4 μm |
| Sterility | Non-sterile |
| Flow Rate | Avg. 49 mL/min x cm2 (typical results @ 10 psi);18 mL/min x cm2/psi |
| For Use With (Application) | Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence. |
| Gas Flow Rate | 10.0 L/min/cm2 |
| Show More |