Learn More
Invitrogen™ SuperScript™ VILO™ cDNA Synthesis Kit
The SuperScript VILO cDNA Synthesis Kit is designed to generate first strand cDNA for two-step RT-qPCR applications. The kit is supplied in a two-tube format with the VILO reaction mix and an enzyme blend in separate tubes.
Supplier: Invitrogen™ 11754050
Description
The SuperScript VILO cDNA Synthesis Kit is designed to generate first strand cDNA for two-step RT-qPCR applications. The kit is supplied in a two-tube format with the VILO reaction mix and an enzyme blend in separate tubes. The enzyme blend contains SuperScript III Reverse Transcriptase (RT), a genetically engineered MMLV RT that has reduced RNase H activity and improved thermostability for highly efficient cDNA synthesis.
The SuperScript VILO cDNA Synthesis Kit can be used to synthesize cDNA from for a wide range of input RNA amounts. This kit provides the high temperature capability of SuperScript III Reverse Transcriptase in an optimized format for generating first-strand cDNA for use in RT–qPCR.
Notes
• For superior first-strand cDNA synthesis in two-step RT-qPCR, try SuperScript IV VILO Master Mix (Cat. No. 11756050) or SuperScript IV VILO Master Mix with ezDNase Enzyme (Cat. No. 11766050).
• This kit is included with EXPRESS Two-Step RT–qPCR kits. Reagents are provided for 50 or 250 20-μL cDNA synthesis reactions.
- Environmental benefits include:
- Renewable Energy
Specifications
• 5X VILO Reaction Mix, 200 μL |
|
Kit | |
60 min. | |
42°C | |
SuperScript III | |
Dry Ice | |
First-Strand cDNA | |
Master Mix | |
Up to 10 kb |
qPCR | |
High | |
Reverse Transcription | |
50 reactions | |
Reduced | |
Real Time PCR (qPCR) | |
50 Reactions | |
Reverse Transcription | |
RNA |
Promotions
-
Thermo Fisher Scientific
,
Applied Biosystems
,
Invitrogen
Get Up to $3,000 of qPCR Products with Purchase
Buy up to $3,000 of qPCR consumables and get a matching amount of qPCR products at no additional cost.Expires: 06/30/26
For Research Use Only. Not for use in diagnostic procedures.