missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ VILO™ cDNA Synthesis Kit

Catalog No. p-4926559
Encompass
Change view
Click to view available options
No. of Reactions:
250 Reactions
50 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11754050 50 Reactions
11754250 250 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11754050 Supplier Invitrogen™ Supplier No. 11754050

Includes

5X VILO Reaction Mix (200μL) and a 10X SuperScript III Enzyme Blend (100μL), which contains: SuperScript III RT, RNaseOUT Recombinant Ribonuclease Inhibitor, and a proprietary helper protein

The SuperScript VILO cDNA Synthesis Kit is designed to generate first strand cDNA for two-step RT-qPCR applications. The kit is supplied in a two-tube format with the VILO reaction mix and an enzyme blend in separate tubes.

The SuperScript VILO cDNA Synthesis Kit is designed to generate first strand cDNA for two-step RT-qPCR applications. The kit is supplied in a two-tube format with the VILO reaction mix and an enzyme blend in separate tubes. The enzyme blend contains SuperScript III Reverse Transcriptase (RT), a genetically engineered MMLV RT that has reduced RNase H activity and improved thermostability for highly efficient cDNA synthesis.

The SuperScript VILO cDNA Synthesis Kit can be used to synthesize cDNA from for a wide range of input RNA amounts. This kit provides the high temperature capability of SuperScript III Reverse Transcriptase in an optimized format for generating first-strand cDNA for use in RT–qPCR.

Notes
• For superior first-strand cDNA synthesis in two-step RT-qPCR, try SuperScript IV VILO Master Mix (Cat. No. 11756050) or SuperScript IV VILO Master Mix with ezDNase Enzyme (Cat. No. 11766050).
• This kit is included with EXPRESS Two-Step RT–qPCR kits. Reagents are provided for 50 or 250 20-μL cDNA synthesis reactions.

Order Info

Shipping Condition: Dry ice

Specifications

Content And Storage

• 5X VILO Reaction Mix, 200 μL
• 10X SuperScript III Enzyme Blend, 100 μL

Store at –5 to –30°C.

Detection Method qPCR
Format Kit
GC-Rich PCR Performance High
Reaction Speed 60 min.
Technique Reverse Transcription
Optimal Reaction Temperature 42°C
Quantity 50 reactions
Reverse Transcriptase SuperScript III
Ribonuclease H Activity Reduced
Shipping Condition Dry Ice
For Use With (Application) Real Time PCR (qPCR)
Final Product Type First-Strand cDNA
No. of Reactions 50 Reactions
Reaction Format Master Mix
Reagent Type Reverse Transcription
Size (Final Product) Up to 10 kb
Starting Material RNA
Show More Show Less
What is the difference between SuperScript III RT and the RT in the SuperScript VILO kit?

The SuperScript VILO cDNA Synthesis Kit contains a mix of SuperScript III RT and helper proteins which help to increase the efficiency of the reverse transcription reaction and thus improve yield. The RT in the SuperScript VILO kit is active at 42 degrees C due to the helper proteins.

How much of the first-strand cDNA reaction should I load for PCR?

While the volume is dependent on the starting amount of RNA used for the first-strand synthesis and the abundance of the target gene, we'd recommend starting with 10% of the first-strand reaction for your PCR reaction.

In what concentration range of total RNA does the SuperScript VILO cDNA Synthesis Kit (Cat. No. 11754050) work?

  • Lower limit of RNA concentration: 1.0 pg
  • Upper limit of RNA concentration: 2.5 µg
  • What is the RNA input for the Oncomine Focus Assay, Chef-Ready Library kit?

    Each reverse transcription reaction requires 10 ng of DNase-treated total RNA (≥1.43 ng/µL).

    With the Oncomine Focus Assay, Chef-Ready Library, can I perform reverse transcription using the SuperScript VILO cDNA Synthesis Kit on the Ion Chef System?

    Reverse transcription needs to be performed manually in the IonCode 96-well PCR plate, using the SuperScript VILO cDNA Synthesis Kit. SuperScript reverse transcriptase enzyme is not supplied in the kit and must be purchased separately. If the RNA was prepared from FFPE tissue and not previously heat-treated, we recommend heating at 80 degrees C for 10 min, then cooling to room temperature.

    For Research Use Only. Not for use in diagnostic procedures.