missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finnpipette™ Novus Multichannel Pipettes
GREENER_CHOICE

Catalog No. p-4530122
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
100 to 1200 μL
30 to 300 μL
5 to 50 μL
Number of Channels:
12
16
8
Color:
Orange
Pink
Turquoise
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels Color
1438601 1 to 10 μL 8 Pink
1438604 1 to 10 μL 12 Pink
1438602 5 to 50 μL 8 Yellow
1438605 5 to 50 μL 12 Yellow
1438603 30 to 300 μL 8 Orange
1438606 30 to 300 μL 12 Orange
1438607 5 to 50 μL 16 Turquoise
21377845 100 to 1200 μL 8 Turquoise
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 1438601 Supplier Thermo Scientific™ Supplier No. 46300000
Only null left

Speed up repetitive dispensing into microplates with these lightweight, multichannel pipettes, with balanced weight distribution for reduced muscle stress and strain.

Speed up repetitive dispensing into 96- and 384-microplates with Thermo Scientific™ Finnpipette™ Novus Multichannel Pipettes. Available in a wide range of models for common laboratory tasks, this product has a lightweight handle and unique index-finger operation that enable longer work periods without fatigue. Simplicity, comfort and functionality — the Novus will be your new best friend in the lab.

Features:

  • Easy-to-read and intuitive menu navigation
  • Multilingual user interface options
  • 10 pipetting options include forward, reverse, diluting, stepper pipetting, mixing, sequential stepping and more
    • 9 aspirate/dispense speeds 
    • Save up to 9 programs with your most common protocols
    • 8 multichannel versions, including a 16-channel for 384-well microplate applications, with color coding by volume for easy recognition
    • Full-size, backlit screen eliminates surrounding light reflections and improves contrast in low-light conditions
    • Quick in-lab calibration for accuracy and precision
    • Fully autoclavable tip cone ensures sterility and avoids cross-contamination
    • Long-life battery allows approximately 4000 pipetting operations; recharged in about one hour

Ergonomic Design

  • Extremely lightweight construction
  • Trigger-action pipetting — index finger controls dispensing with just a soft press of a button
  • 120° adjustable finger rest for balance and comfort
  • Soft-touch tip ejection minimizes thumb strain; can be operated by either hand

Compatible with:

Ideal for use with Thermo Scientific Finntip™ Flex or Finntip pipette tips

Includes:

Finnpipette Novus, universal charger, piston removal tool, instructions for use and calibration certificate

Specifications

Accuracy 12.0% ±2.4
Compatible Tips Flex 10, 10, 20 micro, 50 micro
Autoclavable Autoclavable
Color Pink
For Use With Flex 10, 10 Micro, 20 Micro, 50 Micro
Model Novus
Number of Channels 8
Product Line Novus
Programmability Yes
Sterility Sterile
Type Multi-Channel Pipette
Volume (Metric) 1 to 10 μL
Increments 0.1 μL
Precision 8.0 to 1.6%
Show More Show Less