missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finnpipette™ Novus Multichannel Pipettes
GREENER_CHOICE

Catalog No. 21377845
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
100 to 1200 μL
30 to 300 μL
5 to 50 μL
Number of Channels:
12
16
8
Color:
Orange
Pink
Turquoise
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels Color
21377845 100 to 1200 μL 8 Turquoise
1438601 1 to 10 μL 8 Pink
1438604 1 to 10 μL 12 Pink
1438602 5 to 50 μL 8 Yellow
1438605 5 to 50 μL 12 Yellow
1438603 30 to 300 μL 8 Orange
1438606 30 to 300 μL 12 Orange
1438607 5 to 50 μL 16 Turquoise
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 21377845 Supplier Thermo Scientific™ Supplier No. 46300800

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Speed up repetitive dispensing into microplates with these lightweight, multichannel pipettes, with balanced weight distribution for reduced muscle stress and strain.

Speed up repetitive dispensing into 96- and 384-microplates with Thermo Scientific™ Finnpipette™ Novus Multichannel Pipettes. Available in a wide range of models for common laboratory tasks, this product has a lightweight handle and unique index-finger operation that enable longer work periods without fatigue. Simplicity, comfort and functionality — the Novus will be your new best friend in the lab.

Features:

  • Easy-to-read and intuitive menu navigation
  • Multilingual user interface options
  • 10 pipetting options include forward, reverse, diluting, stepper pipetting, mixing, sequential stepping and more
    • 9 aspirate/dispense speeds 
    • Save up to 9 programs with your most common protocols
    • 8 multichannel versions, including a 16-channel for 384-well microplate applications, with color coding by volume for easy recognition
    • Full-size, backlit screen eliminates surrounding light reflections and improves contrast in low-light conditions
    • Quick in-lab calibration for accuracy and precision
    • Fully autoclavable tip cone ensures sterility and avoids cross-contamination
    • Long-life battery allows approximately 4000 pipetting operations; recharged in about one hour

Ergonomic Design

  • Extremely lightweight construction
  • Trigger-action pipetting — index finger controls dispensing with just a soft press of a button
  • 120° adjustable finger rest for balance and comfort
  • Soft-touch tip ejection minimizes thumb strain; can be operated by either hand

Compatible with:

Ideal for use with Thermo Scientific Finntip™ Flex or Finntip pipette tips

Includes:

Finnpipette Novus, universal charger, piston removal tool, instructions for use and calibration certificate

Specifications

Accuracy 3.0% ±1.0
Compatible Tips Flex 1200
Autoclavable Autoclavable
Color Turquoise
For Use With Flex 1200
Model Novus
Number of Channels 8
Product Line Novus
Programmability Yes
Sterility Sterile
Type Multi-Channel Pipette
Volume (Metric) 100 to 1200 μL
Increments 1 μL
Precision 0.9 to 0.2%
Show More Show Less