Learn More
MilliporeSigma™ Replacement Solutions for Montage™ SEQ96 and SEQ384 Sequencing Reaction Cleanup Kits
provide efficient, centrifugation-free sequencing reaction cleanup in 96 and 384-well formats.
Supplier: MilliporeSigma™ LSKSIS500
Description
Montage SEQ kits provide efficient, centrifugation-free sequencing reaction cleanup in 96 and 384-well formats. The unique size-exclusion separation technology eliminates the need for costly gel filtration resins or bind-wash-elute columns.Specifications
Montage™ SEQ96 and SEQ384 sequencing reaction cleanup kits | |
Plasmid Miniprep, BAC Prep | |
Injection Solution | |
500 mL |