Learn More
MilliporeSigma™ Replacement Solutions for Montage™ SEQ96 and SEQ384 Sequencing Reaction Cleanup Kits
provide efficient, centrifugation-free sequencing reaction cleanup in 96 and 384-well formats.
$209.00 - $231.00
Spécifications
For Use With (Equipment) Montage™ SEQ96 and SEQ384 sequencing reaction cleanup kits
For Use With (Application) Plasmid Miniprep, BAC Prep
Quantity 500 mL
voir les spécifications
Produits 2
Numéro de catalogue Numéro du manufacturier. Product Type Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Product Type Prix Quantité  
LSKSBW500
Afficher les documents
MilliporeSigma™
LSKSBW500
Wash Solution
chaque for $231.00
Il en reste null
LSKSIS500
Afficher les documents
MilliporeSigma™
LSKSIS500
Injection Solution
chaque for $209.00
Il en reste null
Description
Description
Montage SEQ kits provide efficient, centrifugation-free sequencing reaction cleanup in 96 and 384-well formats. The unique size-exclusion separation technology eliminates the need for costly gel filtration resins or bind-wash-elute columns.
Spécifications
Spécifications
Montage™ SEQ96 and SEQ384 sequencing reaction cleanup kits
Plasmid Miniprep, BAC Prep
500 mL
Videos
FDS
missing translation for 'documents'
missing translation for 'documents'