missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Express™ PLUS Membrane Filters

Plain white polyethersulfone Express™ PLUS Membrane Filters provide ultrafast filtration for a variety of applications.

Supplier:  MilliporeSigma™ HPWP01300

 View more versions of this product

Catalog No. HPWP01300


Only null left
Explore available promotions

Description

Description

Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications

The Millipore Express™ PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.

Features and Benefit:
  • Ultra fast filtration and high throughput shorten process time
  • Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications
  • Applications:Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration
Specifications

Specifications

Not Autoclavable
Reusable
Disc
0.45 ÎĽm
23 (at 690 Mbar (10 psi))
Hydrophilic
Hydrophilic
13 mm
White
Screen Filter
PES
Non-sterile
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices.
Filter Membranes
0.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions