missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Express™ PLUS Membrane Filters
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
$144.00 - $617.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | White |
Filter Type | Screen filter |
Material | PES |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Diameter (Metric) Filter | Pore Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Filter | Pore Size | Price | Quantity | ||||
GPWP04700
|
MilliporeSigma™
GPWP04700 |
47 mm | 0.22 μm |
Pack of 100 Each for $243.00
|
|
GPWP01300
|
MilliporeSigma™
GPWP01300 |
13 mm | 0.22 μm |
Pack of 100 Each for $150.00
|
|
GPWP02500
|
MilliporeSigma™
GPWP02500 |
25 mm | 0.22 μm |
Pack of 100 Each for $157.00
|
|
GPWP09050
|
MilliporeSigma™
GPWP09050 |
90 mm | 0.22 μm |
Pack of 50 Each for $274.00
|
|
GPWP14250
|
MilliporeSigma™
GPWP14250 |
142 mm | 0.22 μm |
Pack of 50 Each for $617.00
|
|
HPWP01300
|
MilliporeSigma™
HPWP01300 |
13 mm | 0.45 μm |
Pack of 100 Each for $144.00
|
|
HPWP14250
|
MilliporeSigma™
HPWP14250 |
142 mm | 0.45 μm |
Pack of 50 Each for $589.00
|
|
HPWP09050
|
MilliporeSigma™
HPWP09050 |
90 mm | 0.45 μm |
Pack of 50 Each for $281.00
|
|
HPWP02500
|
MilliporeSigma™
HPWP02500 |
25 mm | 0.45 μm |
Pack of 100 Each for $195.00
|
|
HPWP04700
|
MilliporeSigma™
HPWP04700 |
47 mm | 0.45 μm |
Pack of 100 Each for $245.00
|
|
Description
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
- Hydrophilic, high-throughput, low-protein-binding
- Thickness: ≥150μm and ≤200μm
- Sterilization: by autoclave (121°C at 1 bar), EO or gamma
- Bacterial endotoxins: 0.5 EU/mL
- Gravimetric extractables: <1.5%
- Flowrate: 40mL/min. cm2 at 2 bar, 25°C
Applications:
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices.
Specifications
Not Autoclavable | |
Screen filter | |
Non-sterile | |
Hydrophilic | |
Hydrophilic |
White | |
PES | |
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices. | |
Filter Membranes |