Learn More
MilliporeSigma™ Centricon™ Plus-70 Centrifugal Filters
15 mL to 70 mL volume
$400.00 - $419.00
Specifications
Disposable Single-use
Sterility Non-sterile
Housing Material Regenerated Cellulose (RC)
Type Centrifugal Filters
Volume (Metric) 15 to 70 mL
View More Specs
Products 4
Catalog Number Mfr. No. MWCO Price Quantity  
Catalog Number Mfr. No. MWCO Price Quantity  
UFC701008
View Documents
MilliporeSigma™
UFC701008
10 kDa
Pack of 8 Each for $400.00
Only null left
UFC710008
View Documents
MilliporeSigma™
UFC710008
100 kDa
Pack of 8 Each for $400.00
Only null left
UFC703008
View Documents
MilliporeSigma™
UFC703008
30 kDa
Pack of 8 Each for $400.00
Only null left
09-720-508
View Documents
MilliporeSigma™
UFC700308
3 kDa
Each for $419.00
Only null left
Description
Description
The Centricon™ Plus-70 centrifugal filter is a disposable, single-use device designed for rapid processing of aqueous biological solutions in volumes ranging from 15 to 70 mL. It is compatible with swinging-bucket centrifuges only. The Centricon Plus-70 device can concentrate most 70 mL solutions down to 350 μL in just 25 minutes (some solutions may take slightly longer).•90% typical recovery •50X to 200X concentration •Low hold-up volume •Polypropylene housing minimizes binding •Deadstop prevents spinning to dryness •Ready to use. Nothing to assemble Applications:•Concentrating and desalting chromatography column eluates •Concentrating monoclonal antibodies •Concentrating proteins or viruses from culture supernatants •Clarifying tissue homogenates and cell lysates •Buffer exchange or diafiltration
disposable,single-use,aqueous,solutions,volumes,ranging
Specifications
Specifications
Single-use
Regenerated Cellulose (RC)
15 to 70 mL
Non-sterile
Centrifugal Filters
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'