Learn More
MilliporeSigma™ Centricon™ Plus-70 Centrifugal Filters
Catalog No. ufc710008
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO
UFC710008 100 kDa
UFC701008 10 kDa
UFC703008 30 kDa
09720508 3 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. UFC710008 Supplier MilliporeSigma™ Supplier No. UFC710008
Only null left
15 mL to 70 mL volume
The Centricon™ Plus-70 centrifugal filter is a disposable, single-use device designed for rapid processing of aqueous biological solutions in volumes ranging from 15 to 70 mL. It is compatible with swinging-bucket centrifuges only. The Centricon Plus-70 device can concentrate most 70 mL solutions down to 350 μL in just 25 minutes (some solutions may take slightly longer).•90% typical recovery •50X to 200X concentration •Low hold-up volume •Polypropylene housing minimizes binding •Deadstop prevents spinning to dryness •Ready to use. Nothing to assemble Applications:•Concentrating and desalting chromatography column eluates •Concentrating monoclonal antibodies •Concentrating proteins or viruses from culture supernatants •Clarifying tissue homogenates and cell lysates •Buffer exchange or diafiltration
disposable,single-use,aqueous,solutions,volumes,ranging
Specifications
Disposable Single-use
Material (Membrane) Ultracel-PL
MWCO 100 kDa
Sterility Non-sterile
For Use With (Application) Concentrating and desalting chromatography column eluates; Concentrating monoclonal antibodies; Concentrating proteins or viruses from culture supernatants; Clarifying tissue homogenates and cell lysates; Buffer exchange or diafiltration
Housing Material Regenerated Cellulose (RC)
Includes Polypropylene filtrate collection cup; sample filter cup; concentrate/retentate cup and cap
Type Centrifugal Filters
Volume (Metric) 15 to 70 mL