missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 1.07 at 25°C, Certified, LabChem™
$72.53 - $277.60
Specifications
Chemical Name or Material | Buffer Solution pH 1.07 at 25°C |
---|---|
CAS Max % | 99.3,0.37,0.33 |
Color | Colorless |
CAS | 7447-40-7,7647-01-0 |
Decomposition Information | Chlorine; Thermal decomposition generates: Corrosive vapors |
Specifications
Buffer Solution pH 1.07 at 25°C | |
Colorless | |
Chlorine; Thermal decomposition generates: Corrosive vapors | |
1.07 ±0.02 | |
Passes Test | |
Poly Bottle | |
Traceable to NIST | |
74.55 |
99.3,0.37,0.33 | |
7447-40-7,7647-01-0 | |
1g/mL | |
Liquid | |
KCl | |
1g/mL | |
Soluble in water | |
Certified |