missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 1.07 at 25°C, Certified, LabChem™
Supplier: LabChem LC122002
Specifications
| Buffer Solution pH 1.07 at 25°C | |
| 99.3,0.37,0.33 | |
| Chlorine; Thermal decomposition generates: Corrosive vapors | |
| 1.07 ±0.02 | |
| 1 L | |
| KCl | |
| 1g/mL | |
| Soluble in water | |
| Certified |
| Colorless | |
| 7447-40-7,7647-01-0 | |
| 1g/mL | |
| Liquid | |
| Passes Test | |
| Poly Bottle | |
| Traceable to NIST | |
| 74.55 |