missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 1.07 at 25°C, Certified, LabChem™
Supplier: LabChem LC122001
Description
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Buffer Solution pH 1.07 at 25°C | |
99.3,0.37,0.33 | |
Chlorine; Thermal decomposition generates: Corrosive vapors | |
1.07 ±0.02 | |
500 mL | |
KCl | |
1g/mL | |
Soluble in water | |
Certified |
Colorless | |
7447-40-7,7647-01-0 | |
1g/mL | |
Liquid | |
Passes Test | |
Poly Bottle | |
Traceable to NIST | |
74.55 |