missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis-Tris, ≥99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219454690
Specifications
Bis-Tris | |
6976-37-0 | |
Crystalline Powder | |
C8H19NO5 | |
2, 2-bis(Hydroxymethyl)-2, 2', 2,2-bis(Hydroxymethyl)-2,2',2''-nitrilotriethanol | |
C(CO)N(CCO)C(CO)(CO)CO | |
209.242 | |
CHEBI:41250 | |
≥99% |
Cell culture reagent | |
White | |
500 g | |
MFCD00002853 | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol | |
81462 | |
209.2 |
Chemical Identifiers
6976-37-0 | |
209.242 | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
81462 | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
C8H19NO5 | |
MFCD00002853 | |
2, 2-bis(Hydroxymethyl)-2, 2', 2,2-bis(Hydroxymethyl)-2,2',2''-nitrilotriethanol | |
CHEBI:41250 | |
C(CO)N(CCO)C(CO)(CO)CO |
Safety and Handling
Recommended Storage : Store at Room Temperature (15-30°C).