missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis-Tris, ≥99%, MP Biomedicals™
$91.88 - $1714.26
Chemical Identifiers
| CAS | 6976-37-0 |
|---|---|
| Molecular Formula | C8H19NO5 |
| Molecular Weight (g/mol) | 209.242 |
| MDL Number | MFCD00002853 |
| InChI Key | OWMVSZAMULFTJU-UHFFFAOYSA-N |
| Synonym | 2,2-bis(Hydroxymethyl)-2,2',2''-nitrilotriethanol, 2, 2-bis(Hydroxymethyl)-2, 2' |
| PubChem CID | 81462 |
| ChEBI | CHEBI:41250 |
| IUPAC Name | 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
| SMILES | C(CO)N(CCO)C(CO)(CO)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19454625
|
MP Biomedicals Inc
0219454625 |
25 g |
Each for $91.88
|
|
|||||
|
ICN19454680
|
MP Biomedicals Inc
0219454680 |
100 g |
Each for $240.04
|
|
|||||
|
ICN19454690
|
MP Biomedicals Inc
0219454690 |
500 g |
Each for $982.51
|
|
|||||
|
ICN19454691
|
MP Biomedicals Inc
0219454691 |
1 kg |
Each for $1,714.26
|
|
|||||
Chemical Identifiers
| 6976-37-0 | |
| 209.242 | |
| OWMVSZAMULFTJU-UHFFFAOYSA-N | |
| 81462 | |
| 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
| C8H19NO5 | |
| MFCD00002853 | |
| 2,2-bis(Hydroxymethyl)-2,2',2''-nitrilotriethanol, 2, 2-bis(Hydroxymethyl)-2, 2' | |
| CHEBI:41250 | |
| C(CO)N(CCO)C(CO)(CO)CO |
Specifications
| 6976-37-0 | |
| Crystalline Powder | |
| C8H19NO5 | |
| 2,2-bis(Hydroxymethyl)-2,2',2''-nitrilotriethanol, 2, 2-bis(Hydroxymethyl)-2, 2' | |
| C(CO)N(CCO)C(CO)(CO)CO | |
| 209.242 | |
| CHEBI:41250 | |
| ≥99% |
| White | |
| 25 g | |
| MFCD00002853 | |
| OWMVSZAMULFTJU-UHFFFAOYSA-N | |
| 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol | |
| 81462 | |
| 209.2 | |
| Bis-Tris, Cell culture reagent |
Safety and Handling
Recommended Storage : Store at Room Temperature (15-30°C).