missing translation for 'onlineSavingsMsg'
Learn More
Learn More
J.T. Baker™ BAKERBOND™ PROchievA™ Recombinant Protein A Resin, Affinity Chromatography Columns, Avantor
J.T.BAKER™ BAKERBOND™ PROchievA™ Recombinant Protein A Resin, Affinity Chromatography Columns offer excellent dynamic binding capacity and alkaline stability linked to Avantor's proprietary ligand.
$672.18 - $12517.35
Specifications
Product Type | Affinity Chromatography Column |
---|---|
Binding Capacity | FIO (Target >65 mg/mL) |
Content And Storage | 2°C to 8°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
02-004-665
|
J.T. Baker™
C78911 |
1 x 1 mL Box |
Each for $672.18
|
|
||||||
02-004-666
|
J.T. Baker™
C78907 |
5 x 1 mL Box |
Each for $2,607.38
|
|
||||||
02-004-667
|
J.T. Baker™
C78918 |
1 x 5 mL Box |
Each for $2,816.15
|
|
||||||
02-004-668
|
J.T. Baker™
C78925 |
5 x 5 mL Box |
N/A
|
N/A | ||||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
Description
- The new protein A resin is designed for high-performance in the critical affinity chromatography step used in the purification of mAbs, Fc fusion proteins, and other similar IgG-like molecules.
- Designed and manufactured by Avantor to the high standards established for J.T. Baker™ brand chemicals, BAKERBOND™ PROchievA™ recombinant protein
- Resin can be used with other J.T. Baker™ BAKERBOND™ resins to enhance the separation power of chromatography process steps
- Available in 1 mL and 5 mL lab columns designed for direct connection to many popular chromatography instruments including GE ÃKTA™ and Bio-Rad NGC™
Specifications
Affinity Chromatography Column | |
FIO (Target >65 mg/mL) | |
2°C to 8°C |