Learn More
J.T. Baker™ BAKERBOND™ PROchievA™ Recombinant Protein A Resin, Affinity Chromatography Columns, Avantor
J.T.BAKER™ BAKERBOND™ PROchievA™ Recombinant Protein A Resin, Affinity Chromatography Columns offer excellent dynamic binding capacity and alkaline stability linked to Avantor's proprietary ligand.
Supplier:  J.T. Baker™ C78918
 View more versions of this product
Catalog No. 02-004-667
Only null left
Explore available promotions
Description
Description
  • The new protein A resin is designed for high-performance in the critical affinity chromatography step used in the purification of mAbs, Fc fusion proteins, and other similar IgG-like molecules.
  • Designed and manufactured by Avantor to the high standards established for J.T. Baker™ brand chemicals, BAKERBOND™ PROchievA™ recombinant protein
  • Resin can be used with other J.T. Baker™ BAKERBOND™ resins to enhance the separation power of chromatography process steps
  • Available in 1 mL and 5 mL lab columns designed for direct connection to many popular chromatography instruments including GE ÄKTA™ and Bio-Rad NGC™
Specifications
Specifications
Affinity Chromatography Column
FIO (Target >65 mg/mL)
2°C to 8°C
1 x 5 mL Box
Product Suggestions
Product Suggestions