missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC ORGZN82Z
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Zn | 1000μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Zinc Oil Standard | |
With Certificate of Analysis | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
MFCD00011291 | |
Certified Reference Material | |
[Zn] | |
zinc | |
65.38 | |
23994 | |
Oil/Plasma |
Triple-leached LDPE bottle | |
7440-66-6,8042-47-5 | |
Zn | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
Base Oil 20 | |
Petrochemical Testing | |
50 g | |
CHEBI:30185 |
Chemical Identifiers
7440-66-6 | |
65.38 | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
CHEBI:30185 | |
[Zn] |
Zn | |
MFCD00011291 | |
23994 | |
zinc |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only