Learn More
Cytiva Whatman™ Cellulose Soxhlet Extraction Thimbles, Single 1mm Wall
GREENER_CHOICE
Manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Whatman™ Cellulose Soxhlet Extraction Thimbles are intended for applications including use in air and waste gas analysis.
$205.89 - $646.11
Specifications
Description GE’s Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Used in air and waste gas analysis.
Material Cellulose
For Use With (Application) Air and waste gas analysis
For Use With (Equipment) Soxhlet extractor
Max. Temperature (Metric) 120°C
View More Specs
Products 23
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inside Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inside Price Quantity  
09-656A
View Documents
Cytiva
2800226
65 mm 22 mm
Pack of 25 Each for $205.89
Only null left
09-656B
View Documents
Cytiva
2800228
80 mm 22 mm
Pack of 25 Each for $216.29
Only null left
09-656D
View Documents
Cytiva
2800266
60 mm 26 mm
Pack of 25 Each for $312.66
Only null left
09-656F
View Documents
Cytiva
2800338
80 mm 33 mm
Pack of 25 Each for $344.06
Only null left
09-656H
View Documents
Cytiva
2800432
123 mm 43 mm
Pack of 25 Each for $455.55
Only null left
09-656L
View Documents
Cytiva
2800250
100 mm 25 mm
Pack of 25 Each for $216.29
Only null left
09-656K
View Documents
Cytiva
2800199
90 mm 19 mm
Pack of 25 Each for $205.89
Only null left
09-656G
View Documents
Cytiva
2800339
94 mm 33 mm
Pack of 25 Each for $350.31
Only null left
09-656J
View Documents
Cytiva
2800608
180 mm 60 mm
Pack of 25 Each for $646.11
Only null left
09-656M
View Documents
Cytiva
2800259
90 mm 25 mm
Pack of 25 Each for $216.29
Only null left
09-656N
View Documents
Cytiva
2800300
100 mm 30 mm
Pack of 25 Each for $233.27
Only null left
09-656P
View Documents
Cytiva
2800308
80 mm 30 mm
Pack of 25 Each for $233.27
Only null left
09-656Q
View Documents
Cytiva
2800331
118 mm 33 mm
Pack of 25 Each for $265.82
Only null left
20-653
View Documents
Cytiva
2800105
50 mm 10 mm
Pack of 25 Each for $205.89
Only null left
09-656C
View Documents
Cytiva
2800258
80 mm 25 mm
Pack of 25 Each for $269.93
Only null left
09927228
View Documents
Cytiva
2800282
120 mm 28 mm
Pack of 25 Each for $228.94
Only null left
09927240
View Documents
Cytiva
2800288
80 mm 28 mm
Pack of 25 Each for $233.27
Only null left
09-927-242
View Documents
Cytiva
2800373
130 mm 37 mm
Pack of 25 Each for $315.74
Only null left
09-927-241
View Documents
Cytiva
2800412
123 mm 41 mm
Pack of 25 Each for $313.00
Only null left
09927244
View Documents
Cytiva
2800280
100 mm 28 mm
Pack of 25 Each for $228.94
Only null left
09927676
View Documents
Cytiva
2800260
100 mm 26 mm
Pack of 25 Each for $216.29
Only null left
09927681
View Documents
Cytiva
2800330
100 mm 33 mm
Pack of 25 Each for $265.82
Only null left
09-927-682
View Documents
Cytiva
2800185
55 mm 18 mm
Pack of 25 Each for $205.89
Only null left
Description
Description
Manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Whatman™ Cellulose Soxhlet Extraction Thimbles are intended for applications including use in air and waste gas analysis.
  • Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
  • Suitable for temperatures up to 120°C (248°F)
  • Excellent mechanical strength and retention
  • Wall thickness of standard thimbles is 1mm to support application need
Specifications
Specifications
GE’s Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Used in air and waste gas analysis.
Air and waste gas analysis
120°C
Cellulose
Soxhlet extractor
Extraction Thimble
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'