Learn More
Cytiva Whatman™ Cyclopore™ Polycarbonate Black Membrane Filters
GREENER_CHOICE
Provides a contrast background for epifluorescence and other microscopy applications. Whatman™ Cyclopore™ Polycarbonate Black Membrane Filters improve test sensitivity with low background fluorescence.
$179.27 - $179.27
Specifications
Color Black
Material Polycarbonate
For Use With (Application) For epifluorescence, other microscopy applications requiring contrasting background; To filter sample to use directly for analysis.
No. per Pack 100Pack
Diameter (Metric) 25 mm
View More Specs
Products 1
Catalog Number Mfr. No. Pore Size Price Quantity  
Catalog Number Mfr. No. Pore Size Price Quantity  
09927985
View Documents
Cytiva
70632504
0.4 μm
Pack of 100 Each for $179.27
Only null left
Description
Description
  • Track-etched
  • High sample visibility provided by the black membrane filter surface
  • Negligible absorption and adsorption of filtrate; nonhygroscopic
  • Temperature resistant up to 140°C
  • Low tare weight
Specifications
Specifications
Black
For epifluorescence, other microscopy applications requiring contrasting background; To filter sample to use directly for analysis.
25 mm
Polycarbonate
100Pack
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'