missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 80

Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 1L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g | CAS: 9005-65-6 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | 604.82 g/mol

Supplier:  Thermo Scientific Chemicals 278630025

 View more versions of this product

Catalog No. AC278630025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Greener Choice Claims
  • Environmental benefits include:
    • Free-of a substance that causes environmental harm
    • Non-Toxic
    • Ozone-Safe or Safe for the Ozone Layer
    • Recyclable
    • Recycled Content
    • Renewable Energy
    • Renewable Materials
Specifications

Specifications

Polysorbate 80
Yellow to Amber
1.0800 g/mL
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Glass bottle
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
604.82
Polyoxyethylene(20)sorbitan monooleate
9005-65-6
Indoors +20°C
2.5 L,2.5 L
MFCD00082107
HDTIFOGXOGLRCB-KTKRTIGZNA-N
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
Solubility in water: 100g/L. Other solubilities: 100g/L isopropyl alcohol, 10g/L xylene, insoluble in propylene glycol and mineral oil
Viscous Liquid

Chemical Identifiers

9005-65-6
Polyoxyethylene(20)sorbitan monooleate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only