missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20

CAS: 9005-64-5 | C26H50O10

$88.96 - $1754.09

Chemical Identifiers

CAS 9005-64-5
Molecular Formula C26H50O10
Synonym Polyoxyethylene(20)sorbitan monolaurate
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Type Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Type Price Quantity  
AC233362500
SDS View Documents
Thermo Scientific Chemicals
233362500
250 mL Glass Bottle
Each for $88.96
Only null left
 
AC233360010
SDS View Documents
Thermo Scientific Chemicals
233360010
1 L Glass Bottle
Each for $165.95
Only null left
 
AC233360051
SDS View Documents
Thermo Scientific Chemicals
233360051
5 L Plastic Bottle
Each for $476.44
Only null left
 
AC233360250
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
233360250
25 L Plastic drum
Each for $1,754.09
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9005-64-5
C26H50O10
Polyoxyethylene(20)sorbitan monolaurate

Specifications

9005-64-5
1.1000 g/mL
Glass Bottle
Solubility in water: 100g/L. Other solubilities: 100g/L isopropylalcohol, xylene, insoluble in mineral oil, 10g/L propylene glycol
Liquid
Colorless to Yellow
C26H50O10
250 mL
Polyoxyethylene(20)sorbitan monolaurate
Polysorbate 20
Videos
SDS
Documents

Documents

RUO – Research Use Only