missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSKgel™ G2000PW HPLC Column, 12 μm Particle Size, Sold by MilliporeSigma™ Supelco™
Use the TSKgel™ G2000PW HPLC column for analysis of water-soluble polymers with molecular weights up to 2,000 Da.
Supplier: Merck Emd Millipore 805105
Description
Order Info
Shipped in water.
Greener Choice Claims
- Environmental benefits include:
- Recycled Content
- Free-of a substance that causes environmental harm
Specifications
7.5 mm | |
60 cm | |
12 μm | |
125 Å | |
80°C | |
Hydroxylated Methacrylate | |
Polymer Particle Platform | |
Stainless Steel Hardware |
1.2 mL/min. | |
40 bar | |
2.0 - 12 | |
≤20% organic solvent | |
L25, L38, L39 | |
TSKgel | |
Size Exclusion |
TSKgel is a registered trademark of Tosoh Corporation